Skip to Content
Merck
All Photos(5)

Key Documents

P4006

Sigma-Aldrich

Phosphotungstic acid hydrate

reagent grade

Synonym(s):

Tungstophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3[P(W3O10)4] · xH2O
CAS Number:
Molecular Weight:
2880.05 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

reagent grade

Quality Level

form

powder

mp

~95 °C (lit.)

SMILES string

O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O

InChI

1S/H3O4P.H2O.36O.12W/c1-5(2,3)4;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/h(H3,1,2,3,4);1H2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;

InChI key

AVFBYUADVDVJQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphotungstic acid hydrate is a heteropolyacid.

Application

Phosphotungstic acid hydrate may be used as a staining agent for the transmission electron microscope (TEM) imaging of cetyltrimethylammonium bromide (CTAB) coated gold nanorods.
It may be used in the preparation of:
  • Phosphotungstic acid doped ormosil hybrid networks.
  • Niobia supported phosphotungstic acid (PTA) catalysts.
  • Titanium dioxide (TiO2) nanoparticles /phosphotungstic acid (HPW) films.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shreyak Chaplot et al.
Journal of food protection, 82(5), 878-888 (2019-04-25)
Atmospheric cold plasma and peracetic acid-based hurdle approach for safety of poultry products was evaluated. Study demonstrates a significant synergetic approach to reducing Salmonella on raw poultry. Hurdle approach shows promising bacterial reduction but requires further optimization.
Competitive Reaction Pathway for Site-Selective Conjugation of Raman Dyes to Hotspots on Gold Nanorods for Greatly Enhanced SERS Performance.
Huang H, et al.
Small, 10(19), 4012-4019 (2014)
Fabrication and characterization of heteropolyacid (H3PW12O40)/directly polymerized sulfonated poly (arylene ether sulfone) copolymer composite membranes for higher temperature fuel cell applications.
Kim YS, et al.
Journal of Membrane Science , 212(1), 263-282 (2003)
Photoactive layer-by-layer films of cellulose phosphate and titanium dioxide containing phosphotungstic acid.
Ullah S, et al.
Applied Surface Science, 277, 111-120 (2013)
Tanja Schulz-Mirbach et al.
Frontiers in zoology, 10(1), 63-63 (2013-10-29)
While a number of studies have illustrated and analyzed 3D models of inner ears in higher vertebrates, inner ears in fishes have rarely been investigated in 3D, especially with regard to the sensory epithelia of the end organs, the maculae.

Articles

Discover the detection of milk adulteration by whey using spectrophotometric estimation of sialic acid in three different milk brands. Learn more about this essential technique in our application note.

Discover the detection of milk adulteration by whey using spectrophotometric estimation of sialic acid in three different milk brands. Learn more about this essential technique in our application note.

Discover the detection of milk adulteration by whey using spectrophotometric estimation of sialic acid in three different milk brands. Learn more about this essential technique in our application note.

Discover the detection of milk adulteration by whey using spectrophotometric estimation of sialic acid in three different milk brands. Learn more about this essential technique in our application note.

Protocols

Enzymatic Assay of Neuraminidase applies to products that have a specification for neuraminidase content by enzymatic determination.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service