Skip to Content
Merck
All Photos(2)

Key Documents

79255

Supelco

Phthalic acid

analytical standard

Synonym(s):

1,2-Benzenedicarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2H)2
CAS Number:
Molecular Weight:
166.13
Beilstein:
608199
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)
≥99.5% (T)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

mp

210-211 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC(C1=C(C(O)=O)C=CC=C1)=O

InChI

1S/C8H6O4/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H,9,10)(H,11,12)

InChI key

XNGIFLGASWRNHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phthalic acid is an organic acid and a major bioactive component, reportedly identified in the extracts of Solanum nigrum,[1] Nigella sativa,[2] Angelicae Radix and Ligustici Rhizoma,[3] utilized as Chinese herbal medicines.

Application

Phthalic acid may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Angelicae Radix and Ligustici Rhizoma using high performance liquid chromatography with photodiode array detection (HPLC-PDA) followed by liquid chromatography coupled to mass spectrometry (LC-MS) and capillary electrophoresis (CE) analyses.[3]
  • Nigella sativa[2] and Solanum nigrum[1] using gas chromatography-mass spectrometry (GC-MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

334.4 °F

Flash Point(C)

168 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation and identification of the organic acids in Angelicae Radix and Ligustici Rhizoma by HPLC and CE
Huang Y-W and Sheu J-S
Journal of Separation Science, 29(17), 2616-2624 (2006)
Characterization of alkaloid constitution and evaluation of antimicrobial activity of Solanum nigrum using gas chromatography mass spectrometry (GC-MS)
Jasim H, et al.
Journal of pharmacognosy and phytotherapy, 7(4), 56-72 (2015)
Analysis of bioactive chemical compounds of Nigella sativa using gas chromatography-mass spectrometry
Hadi YM, et al.
Journal of pharmacognosy and phytotherapy, 8(2), 8-24 (2016)
En-Cui Yang et al.
Dalton transactions (Cambridge, England : 2003), 44(7), 3190-3199 (2015-01-13)
Four new 4-amino-1,2,4-triazole (atr)-based coordination polymers, {[Cu2(atr)(H2O)(μ-OH)2(pa)]·H2O}n (), {[Cu3(atr)2(H2O)2(μ-OH)2(npa)2]·2H2O}n (), {[Cu3(atr)5(dca)(μ-OH)(ClO4)2](ClO4)2}n () and {[Cu3(atr)2(H2O)(μ3-OH)(μ-OH)2(spa)]·1.5H2O}n () (pa(2-) = phthalate, npa(2-) = 3-nitrophthalate, dca(-) = dicyanamide and spa(3-) = 4-sulfophthalate), were successfully obtained by varying the carboxylate- and cyanide-modified magnetic bridges. Structural
Germaine M Buck Louis et al.
Fertility and sterility, 101(5), 1359-1366 (2014-02-19)
To assess the relationship between environmental chemicals and couple fecundity or time to pregnancy (TTP). Prospective cohort. Communities of targeted populations with reported exposure. 501 couples recruited upon discontinuing contraception to become pregnant, 2005-2009. None. Fecundability odds ratios (FORs) and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service