Skip to Content
Merck
All Photos(1)

Key Documents

796832

Sigma-Aldrich

Phthalic acid mono-2-ethylhexyl ester

97%

Synonym(s):

mono-2-Ethylhexyl phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O4
CAS Number:
Molecular Weight:
278.34
Beilstein:
3206630
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

description

Flash Point: >110°C

Assay

97%

form

liquid (Colorless)

refractive index

n/D 1.5051

density

1.0864 g/mL at 25 °C

functional group

carboxylic acid
ester

SMILES string

O=C(O)C1=CC=CC=C1C(OCC(CC)CCCC)=O

InChI

1S/C16H22O4/c1-3-5-8-12(4-2)11-20-16(19)14-10-7-6-9-13(14)15(17)18/h6-7,9-10,12H,3-5,8,11H2,1-2H3,(H,17,18)

InChI key

DJDSLBVSSOQSLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phthalic acid mono-2-ethylhexyl ester can be used as a precursor:
  • To synthesize dioctyl phthalate, a widely used plasticizer, via esterification reaction with 2-ethylhexanol.[1]
  • To prepare a fluorescent probe, 2-[[(3′,6′-dihydroxy-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthen]-5-yl)amino]carbonyl] 2-ethylhexyl benzoic acid ester, to detect toxic metabolite mono-2-ethylhexyl phthalate (MEHP).[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunjiao Lu et al.
Ecotoxicology and environmental safety, 208, 111525-111525 (2020-10-30)
The base excision repair (BER) pathway is an important defense response to oxidative DNA damage. It is known that exposures to phthalate esters (PAEs), including Dibutyl phthalate (DBP), Mono-(2-ethylhexyl) phthalate (MEHP), and Di-(2-ethylhexyl) phthalate (DEHP), cause reactive oxygen species-induced DNA
In Situ Fluorescence Tracking Toxic Metabolite Mono-2-ethylhexyl phthalate (MEHP) of Di-(2-ethylhexyl) phthalate (DEHP) in HeLa Cells
Huo Shao-Jie, et al.
Chemical Research in Toxicology, 32(10), 2006-2015 (2019)
Reactive Extraction Strategy for Synthesizing Dioctyl Phthalate Intensified by Bifunctional Deep Eutectic Solvent [Im: 2PTSA]
Qin Hao, et al.
Chemical Engineering and Processing, 157, 108060-108060 (2020)
Da-Hye Kim et al.
Molecules (Basel, Switzerland), 24(8) (2019-04-24)
Endocrine active compounds with structural similarities to natural hormones such as 17β-estradiol (E2) and androgen are suspected to affect the human endocrine system by inducing hormone-dependent effects. This study aimed to detect the (anti-)estrogenic and (anti-)androgenic activities of mono-(2-ethylhexyl) phthalate
Tong-Dian Zhang et al.
Oxidative medicine and cellular longevity, 2020, 4569268-4569268 (2020-06-23)
Mono-(2-ethylhexyl) phthalate (MEHP) and genistein have been classified as endocrine-disrupting chemicals (EDCs) which interfere with the differentiation and development of the male reproductive system. However, how these two EDCs would affect fetal rat testis development at a low dose was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service