Skip to Content
Merck
All Photos(1)

Documents

12540

Supelco

Benzene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11
Beilstein:
969212
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.77 (vs air)

vapor pressure

166 mmHg ( 37.7 °C)
74.6 mmHg ( 20 °C)

Assay

≥99.9% (GC)

autoignition temp.

1043 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.03% water
water

refractive index

n20/D 1.501 (lit.)
n20/D 1.501

bp

80 °C (lit.)

mp

5.5 °C (lit.)

density

0.874 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

c1ccccc1

InChI

1S/C6H6/c1-2-4-6-5-3-1/h1-6H

InChI key

UHOVQNZJYSORNB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Benzene may be used as a reference standard in the determination of the analyte in beverages using headspace gas chromatography coupled with flame ionization detector (HS-GC-FID).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B - Skin Irrit. 2 - STOT RE 1

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

12.2 °F

Flash Point(C)

-11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Validation and uncertainty estimation of analytical method for determination of benzene in beverages.
Styarini D, et al.
Eurasian Journal of Analytical Chemistry, 6(3), 159-172 (2011)
David Galbraith et al.
Critical reviews in toxicology, 40 Suppl 2, 1-46 (2010-10-14)
Over the last century, benzene has been a well-studied chemical, with some acute and chronic exposures being directly associated with observed hematologic effects in humans and animals. Chronic heavy exposures to benzene have also been associated with acute myelogenous leukemia
Audrey Smargiassi et al.
Environmental research, 132, 38-45 (2014-04-20)
The acute cardiorespiratory effects of air quality among children living in areas with considerable heavy industry have not been well investigated. We conducted a panel study of children with asthma living in proximity to an industrial complex housing two refineries
Jelle Vlaanderen et al.
Environmental health perspectives, 119(2), 159-167 (2010-10-01)
The use of occupational cohort studies to assess the association of benzene and lymphoma is complicated by problems with exposure misclassification, outcome classification, and low statistical power. We performed meta-analyses of occupational cohort studies for five different lymphoma categories: Hodgkin
Cliona M McHale et al.
Carcinogenesis, 33(2), 240-252 (2011-12-15)
Benzene causes acute myeloid leukemia and probably other hematological malignancies. As benzene also causes hematotoxicity even in workers exposed to levels below the US permissible occupational exposure limit of 1 part per million, further assessment of the health risks associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service