Skip to Content
Merck
All Photos(1)

Key Documents

58958

Supelco

Ethyl acetate

analytical standard

Synonym(s):

EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3 (20 °C, vs air)

vapor pressure

73 mmHg ( 20 °C)

Assay

≥99.9% (GC)

autoignition temp.

801 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

2.2-11.5 %, 38 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

solubility

water: soluble

density

0.902 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl acetate may be used as an extractant solvent in the analysis of moxalactum in serum samples using high performance liquid chromatography (HPLC).
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maher F J and Winchester F J.
Frontiers in peritoneal dialysis (2013)
K J S Anand et al.
British journal of anaesthesia, 101(5), 680-689 (2008-08-30)
Relationships between plasma morphine concentrations and neonatal responses to endotracheal tube (ETT) suctioning are unknown in preterm neonates. Ventilated preterm neonates (n=898) from 16 centres were randomly assigned to placebo (n=449) or morphine (n=449). After an i.v. loading dose (100
Jessica Le Ven et al.
Journal of mass spectrometry : JMS, 47(11), 1500-1509 (2012-11-14)
Annonaceous acetogenins (AAGs) are a homogenous class of polyketides proposed as environmental neurotoxins. Previous dereplication studies of AAGs were limited by the use of low-resolution mass spectrometers. Only poor information in terms of structures was provided due to the limited
Yuusuke Fujimoto et al.
PloS one, 7(11), e48685-e48685 (2012-11-13)
Combination therapy with ribavirin, interferon, and viral protease inhibitors could be expected to elicit a high level of sustained virologic response in patients infected with hepatitis C virus (HCV). However, several severe side effects of this combination therapy have been
Flavio G Rocha et al.
Biomaterials, 29(19), 2884-2890 (2008-04-09)
Our group has previously created a functional neointestine that is capable of restoring absorptive function. However, the endogenous level of vascular endothelial growth factor (VEGF) is markedly reduced in the construct compared to native bowel. Therefore, we wanted to locally

Articles

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service