Skip to Content
Merck
All Photos(1)

Key Documents

78445

Supelco

Tetrahydrofuran

analytical standard

Synonym(s):

Butylene oxide, Oxolane, Tetramethylene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
Beilstein:
102391
EC Number:
MDL number:
UNSPSC Code:
12191501
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.5 (vs air)

vapor pressure

114 mmHg ( 15 °C)
143 mmHg ( 20 °C)

Assay

≥99.9% (GC)

autoignition temp.

610 °F

shelf life

limited shelf life, expiry date on the label

contains

~0.025% 2,6-di-tert.-butyl-4-methylphenol as stabilizer

expl. lim.

1.8-11.8 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.407 (lit.)

bp

65-67 °C (lit.)

mp

−108 °C (lit.)

solubility

H2O: soluble

density

0.889 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

C1CCOC1

InChI

1S/C4H8O/c1-2-4-5-3-1/h1-4H2

InChI key

WYURNTSHIVDZCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tetrahydrofuran (THF) is a saturated cyclic ether. Its potential use as a biofuel for internal combustion engines is investigated.
Tetrahydrofuran is an important basic chemical compound, which belongs to the category of 5-membered heterocycles.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tetrahydrofuran may be used as an extraction solvent for the determination of pesticide residues in olive oil and phthalates in toys and child care articles by chromatography techniques.
Tetrahydrofuran may find application as a solvent in the synthesis of (S)-2-(1-pyrrolidinylmethyl)pyrrolidine. It is also an excellent solvent for sodium diisopropylamide, which can find applications as an effective base for the isomerization of alkenes and metalation of 1,4-dienes.

recommended

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-6.2 °F - closed cup

Flash Point(C)

-21.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct Functionalization of Tetrahydrofuran and 1, 4-Dioxane: Nickel-Catalyzed Oxidative C (sp3) H Arylation
Liu D, et al.
Angewandte Chemie (International Edition in English), 52(16), 4453-4456 (2013)
Hayashi T, et al. et al.
Asymmetric Synthesis: Graphical Abstracts and Experimental Methods (1998)
Sodium Diisopropylamide in Tetrahydrofuran: Selectivities, Rates, and Mechanisms of Alkene Isomerizations and Diene Metalations
Algera FR, et al.
Journal of the American Chemical Society, 139(33), 11544- 11549 (2017)
Simultaneous GC?MS determination of eight phthalates in total and migrated portions of plasticized polymeric toys and childcare articles
Al-Natsheh M, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 985, 103-109 (2015)
Determination of residual pesticides in olive oil by GC-MS and HPLC-MS after extraction by size-exclusion chromatography.
Barrek S
Analytical and Bioanalytical Chemistry, 376(3), 355-359 (2003)

Articles

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Learn about analyzing organic volatile impurities (OVIs) in pharmaceuticals using SH-GC, focusing on suitable solvents and ensuring compliance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service