Skip to Content
Merck
All Photos(1)

Documents

PHR1084

Supelco

Metformin hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1,1-Dimethylbiguanide hydrochloride, Metformin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(=NH)NHC(=NH)N(CH3)2 · HCl
CAS Number:
Molecular Weight:
165.62
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. M0605000
traceable to USP 1396309

API family

metformin

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

223-226 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CN(C)C(=N)NC(N)=N

InChI

1S/C4H11N5.ClH/c1-9(2)4(7)8-3(5)6;/h1-2H3,(H5,5,6,7,8);1H

InChI key

OETHQSJEHLVLGH-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Metformin is a biguanide hypoglycaemic agent and an oral antidiabetic medication widely used in the treatment of type 2 diabetes. It is effective in glycemic control and decreases the intestinal absorption of glucose, without the problem of serious lactic acidosis.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Metformin hydrochloride may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using different chromatography techniques.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Metformin is an antidiabetic agent that reduces blood glucose levels and improves insulin sensitivity. Its metabolic effects, including the inhibition of hepatic gluconeogenesis, are mediated at least in part by activation of the LKB1-AMPK (AMP-activated protein kinase) pathway. Activation of this pathway also appears to be involved in the antiproliferative and proapoptotic actions of metformin in cancer cell lines.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous estimation of metformin hydrochloride and pioglitazone hydrochloride by RPHPLC method from combined tablet dosage form
Sahoo KP, et al.
Indian Journal of Pharmaceutical Sciences, 70(3), 383-383 (2008)
Simultaneous determination of metformin and pioglitazone by reversed phase HPLC in pharmaceutical dosage forms
Lakshmi SK, et al.
International Journal of Pharmacy and Pharmaceutical Sciences, 1(2), 162-166 (2009)
Stability-indicating HPTLC method for simultaneous determination of nateglinide and metformin hydrochloride in pharmaceutical dosage form
Thomas BA, et al.
Saudi Pharmaceutical Journal : SPJ : The Official Publication of the Saudi Pharmaceutical Society, 19(4), 221-231 (2011)
Qian Zhao et al.
Biochemical pharmacology, 156, 444-450 (2018-09-18)
Metformin, a commonly used agent in the treatment of type 2 diabetes, is also associated with reduced risk of cancer development and improvement in cancer survival. Although much is known about metformin, the mechanisms behind its anti-cancer properties are not
T W Hale et al.
Diabetologia, 45(11), 1509-1514 (2002-11-19)
The aim of this study was to characterize the milk-to-plasma ratio and infant dose for metformin in breastfeeding women, and to measure plasma concentrations and assess any effects in their infants. We hypothesized that metformin used by mothers is safe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service