Skip to Content
Merck
All Photos(2)

Documents

56226

Supelco

Rhodamine 6G

analytical standard

Synonym(s):

Basic Red 1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C28H31N2O3Cl
CAS Number:
Molecular Weight:
479.01
Colour Index Number:
45160
Beilstein:
3900071
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Cl.CCNc1cc2OC3=CC(=N/CC)\C(C)=CC3=C(c2cc1C)c4ccccc4C(=O)OCC

InChI

1S/C28H30N2O3.ClH/c1-6-29-23-15-25-21(13-17(23)4)27(19-11-9-10-12-20(19)28(31)32-8-3)22-14-18(5)24(30-7-2)16-26(22)33-25;/h9-16,29H,6-8H2,1-5H3;1H/b30-24+;

InChI key

VYXSBFYARXAAKO-WTKGSRSZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

A rhodamine analog useful in Pgp efflux assays. Has been used to characterize kinetics of MRP1- mediated efflux.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asmahan Abu-Arish et al.
Biophysical journal, 97(8), 2164-2178 (2009-10-22)
The GTPase Ran is a key regulator of molecular transport through nuclear pore complex (NPC) channels. To analyze the role of Ran in its nuclear transport function, we used several quantitative fluorescence techniques to follow the distribution and dynamics of
Christian Eggeling et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 6(5), 791-804 (2005-05-11)
Under high-excitation irradiance conditions in one- and two-photon induced fluorescence microscopy, the photostability of fluorescent dyes is of crucial importance for the detection sensitivity of single molecules and for the contrast in fluorescence imaging. Herein, we report on the dependence
Xiaobin Xu et al.
Advanced materials (Deerfield Beach, Fla.), 24(40), 5457-5463 (2012-08-14)
Surface enhanced Raman scattering (SERS) is sensitive enough for single-molecule biochemical detection, but it is extremely difficult to obtain a large number of SERS hotspots for sensitive and reproducible detection. It is even more challenging to assemble the hotspots at
Ping Wu et al.
Analytical chemistry, 84(18), 7692-7699 (2012-08-29)
The aptamer (S2.2)-guided Ag-Au nanostructures (aptamer-Ag-Au) have been synthesized by photoreduction and validated by ultraviolet-visible light (UV-vis) spectra and transmission electron microscopy (TEM) images. Differential interference contrast (DIC), fluorescence, and TEM images, and surface-enhanced Raman scattering (SERS) spectra indicated that
M Ursula Norman et al.
The American journal of pathology, 172(6), 1638-1649 (2008-05-10)
The role that mast cells play during contact hypersensitivity (CS) response is unclear because some studies have shown that mast cell-deficient mice have relatively intact CS responses whereas others have shown opposing results. Mast cells secrete a wide range of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service