Skip to Content
Merck
All Photos(1)

Key Documents

R4127

Sigma-Aldrich

Rhodamine 6G

Dye content ~95 %, Powder

Synonym(s):

Basic Red 1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C28H31N2O3Cl
CAS Number:
Molecular Weight:
479.01
Colour Index Number:
45160
Beilstein:
3900071
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Rhodamine 6G, Dye content ~95 %

form

powder

composition

Dye content, ~95%

technique(s)

titration: suitable

mp

215 °C

solubility

H2O: 1 mg/mL (Red to Very Dark Red)

density

1.3 g/cm3

εmax

≥1700 at 525 nm in water

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.CCNc1cc2OC3=CC(=N/CC)\C(C)=CC3=C(c2cc1C)c4ccccc4C(=O)OCC

InChI

1S/C28H30N2O3.ClH/c1-6-29-23-15-25-21(13-17(23)4)27(19-11-9-10-12-20(19)28(31)32-8-3)22-14-18(5)24(30-7-2)16-26(22)33-25;/h9-16,29H,6-8H2,1-5H3;1H/b30-24+;

InChI key

VYXSBFYARXAAKO-WTKGSRSZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Rhodamine 6G has been used as a leukocyte marker to study the interaction between leukocytes and the vascular endothelium. It has also been used to label platelets.

Biochem/physiol Actions

A rhodamine analog useful in Pgp efflux assays. Has been used to characterize kinetics of MRP1- mediated efflux.
Rhodamine 6G is a rhodamine analog useful in P-glycoprotein (Pgp) efflux assays. It has been used to characterize kinetics of multidrug resistance protein 1 (MRP1)-mediated efflux.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interaction of Kupffer Cells and Platelets Determines the Severity of Ischemia-Reperfusion Injury in Steatosis
Koichi Ogawa
The Tohoku Journal of Experimental Medicine, 232 (2014)
Discoidin Domain Receptor 1 Is a Major Mediator of Inflammation and Fibrosis in Obstructive Nephropathy
Dominique G
The American Journal of Pathology (2011)
Şefik E Erdener et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 41(2), 236-252 (2020-04-03)
Ever since the introduction of thrombolysis and the subsequent expansion of endovascular treatments for acute ischemic stroke, it remains to be identified why the actual outcomes are less favorable despite recanalization. Here, by high spatio-temporal resolution imaging of capillary circulation
Kinetic Analysis of Rhodamines Efflux Mediated by the Multidrug Resistance Protein (MRP1)
Chantarawan S
Biophysical Journal, 85(3) (2003)
Impact of pore size on the vascularization and
osseointegration of ceramic bone substitutes in vivo
Frank M. Klenke
Journal of Biomedical Materials Research (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service