Skip to Content
Merck
All Photos(3)

Key Documents

R8004

Sigma-Aldrich

Rhodamine 123

Powder

Synonym(s):

2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C21H17ClN2O3
CAS Number:
Molecular Weight:
380.82
Beilstein:
6030951
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Rhodamine 123, mitochondrial specific fluorescent dye

form

powder

solubility

ethanol: 1 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl[H].COC(=O)c1ccccc1C2=C3C=CC(=N)C=C3Oc4cc(N)ccc24

InChI

1S/C21H16N2O3.ClH/c1-25-21(24)15-5-3-2-4-14(15)20-16-8-6-12(22)10-18(16)26-19-11-13(23)7-9-17(19)20;/h2-11,22H,23H2,1H3;1H

InChI key

MYFATKRONKHHQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Rhodamine 123 is useful as a laser dye and to have selective cell growth effects. It has also been used to measure mitochondrial membrane potential.

Biochem/physiol Actions

Fluorescent dye most commonly used in flow cytometry as functional reporter for Pgp. Functional assays for MDR proteins are better prognostic indicators in cancer therapy than levels of MDR protein expression. Rh 123 can be used in multiparameter analyses without fluorescence interference in combination with common protein labeling dyes such as PE-Cy5 and AMCA (7-amino-4-methylcoumarin-3-acetic acid). Recent reports indicate Rh 123 may also be a substrate of MRP1.
Rhodamine 123 is a fluorescent dye most commonly used in flow cytometry as functional reporter for P-glycoprotein (Pgp). It is part of the rhodamine family of flurone dyes. The dye is mostly used to assess mitochondrial bioenergetics in living cells. Functional assays for multi-drug resistance (MDR) proteins are better prognostic indicators in cancer therapy than levels of MDR protein expression. Rh 123 can be used in multiparameter analyses without fluorescence interference in combination with common protein labeling dyes such as PE-Cy5 and AMCA (7-amino-4-methylcoumarin-3-acetic acid). Recent reports indicate Rh 123 may also be a substrate of multi-drug resistance protein-1 (MRP1).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Bernal et al.
Science (New York, N.Y.), 218(4577), 1117-1119 (1982-12-10)
Rhodamine-123, a cationic laser dye, markedly reduced the clonal growth of carcinoma cells but had little effect on nontumorigenic epithelial cells in vitro. This selective inhibitory effect of Rhodamine-123 on some carcinomas is unusual since known anticancer drugs, such as
Advances in Pharmacology (1999)
Paul E Abraham et al.
The Plant journal : for cell and molecular biology, 103(2), 869-888 (2020-04-22)
Crassulacean acid metabolism (CAM) is a specialized mode of photosynthesis that offers the potential to engineer improved water-use efficiency (WUE) and drought resilience in C3 plants while sustaining productivity in the hotter and drier climates that are predicted for much
Characterization of Rhodamine-123 as a Tracer Dye for Use In In vitro Drug Transport Assays
Samantha
PLoS ONE (2012)
BAG5 Interacts with DJ-1 and Inhibits the NeuroprotectiveEffects of DJ-1 to Combat Mitochondrial Oxidative Damage
Li-xia Qin
Oxidative Medicine and Cellular Longevity (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service