Skip to Content
Merck
All Photos(1)

Key Documents

45788

Supelco

9,10-Diphenylanthracene

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H18
CAS Number:
Molecular Weight:
330.42
Beilstein:
1914010
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

245-248 °C (lit.)

application(s)

environmental

format

neat

SMILES string

c1ccc(cc1)-c2c3ccccc3c(-c4ccccc4)c5ccccc25

InChI

1S/C26H18/c1-3-11-19(12-4-1)25-21-15-7-9-17-23(21)26(20-13-5-2-6-14-20)24-18-10-8-16-22(24)25/h1-18H

InChI key

FCNCGHJSNVOIKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

9,10-Diphenylanthracene, an aromatic hydrocarbon, is a blue light emitting material that is used for the measurement of fluorescence quantum yields in dilute solutions. Its derivatives show potential candidature in organic light emitting diode (OLED) devices.[1][2] It is also used as a fluorophore for the study of peroxyoxalate chemiluminescence (POCL).[3][4]

Application

9,10-Diphenylanthracene may be used as an analytical standard for the determination of the analyte in fish and meat products by liquid chromatography (LC) as well as capillary gas chromatography-mass spectrometry (GC-MS) techniques.[5] It may be used as an internal standard for the determination of halofantrine and desbutylhalofantrine in animal biological samples using on-line UV irradiation and POCL detection.[6]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of polycyclic aromatic hydrocarbons in some Canadian commercial fish, shellfish, and meat products by liquid chromatography with confirmation by capillary gas chromatography-mass spectrometry
Lawrence JF and Weber DF
Journal of Agricultural and Food Chemistry, 32(4), 789-794 (1984)
High efficient organic light emitting diodes using new 9, 10-diphenylanthracene derivatives containing bulky substituents on 2, 6-positon
Jo WJ, et al.
Synthetic Metals, 159(13), 1359-1364 (2009)
Analytical applications of peroxyoxalate chemiluminescence
Tsunoda M and Imai K
Analytica Chimica Acta, 541(1-2), 13-23 (2005)
Spectral and photophysical properties of 9, 10-diphenylanthracene in perfluoro-n-hexane: the influence of solute?solvent interactions
Maciejewski A and Steer RP
Journal of Photochemistry, 35(1), 59-69 (1986)
Abena Amponsaa-Karikari et al.
Biomedical chromatography : BMC, 23(1), 101-106 (2008-10-01)
A sensitive, selective and reliable method has been developed and validated for the determination of halofantrine and its metabolite desbutylhalofantrine in rat plasma using 9,10-diphenylanthracene as an internal standard. The method is based on peroxyoxalate chemiluminescence detection of hydrogen peroxide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service