Skip to Content
Merck
All Photos(2)

Key Documents

30181

Supelco

Fluorescein sodium salt

analytical standard

Synonym(s):

Acid Yellow 73, D&C;Yellow No. 8, NaFl, NaFluo, Sodium fluorescein, Uranine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10Na2O5
CAS Number:
Molecular Weight:
376.27
Colour Index Number:
45350
Beilstein:
3833041
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤15% water

fluorescence

λex 460 nm; λem 515 nm(lit.)

format

neat

SMILES string

[Na+].[Na+].[O-]c1ccc2c(Oc3cc([O-])ccc3C24OC(=O)c5ccccc45)c1

InChI

1S/C20H12O5.2Na/c21-11-5-7-15-17(9-11)24-18-10-12(22)6-8-16(18)20(15)14-4-2-1-3-13(14)19(23)25-20;;/h1-10,21-22H;;/q;2*+1/p-2

InChI key

RGPLGPBQJOQFJS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescein sodium salt (uranine) is a xanthene dye, which finds applications as a fluorescent tracer and shows the best of fluorescence in the pH range 6 to 11.[1] Uranine is also employed as a coloring additive in drugs and cosmetics. It is known to exhibit optical chemical sensing and fluorescent properties in biological staining procedures.[2]

Application

Fluorescein sodium salt is used as an analytical standard in determining the formation of hydroxy radicals, by the ZnO-based photocatalysts under UV-filtered simulated sunlight using spectrofluorometry.[3] It is also used as a calibration standard for the label-free measurements of metabolic events within cells in the living animals using fluorescence lifetime imaging microscopy (FLIM).[4]
It may also be used as an analytical reference standard for the quantification of the analyte in the following:
  • Soil samples using high-performance liquid chromatography (HPLC).[2]
  • Coffee samples using high-performance liquid chromatography (HPLC) and capillary electrophoresis (CZE).[5]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analysis Note

remarks on ICP: Na positive

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

423.7 °F - Pensky-Martens closed cup

Flash Point(C)

217.6 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Method for the analysis of phloxine B, uranine, and related xanthene dyes in soil using supercritical fluid extraction and high-performance liquid chromatography
Alcantara-Licudine PJ, et al.
Journal of Agricultural and Food Chemistry, 45(3), 766-773 (1997)
Highly reduced ecotoxicity of ZnO-based micro/nanostructures on aquatic biota: Influence of architecture, chemical composition, fixation, and photocatalytic efficiency
Serra A, et al.
Water Research, 169(9), 115210-115210 (2020)
Phytochemical Characterization of Rhus coriaria L. Extracts by Headspace Solid-Phase Micro Extraction Gas Chromatography, Comprehensive Two-Dimensional Liquid Chromatography, and Antioxidant Activity Evaluation
Arena K, et al.
Molecules (Basel), 27(5), 1727-1727 (2022)
Analysis of phloxine B and uranine in coffee by high-performance liquid chromatography and capillary zone electrophoresis after solid phase extraction cleanup
Alcantara-Licudine PJ, et al.
Journal of Agricultural and Food Chemistry, 46(3), 1005-1011 (1998)
Solid-phase extraction (SPE) of the fluorescence tracers uranine and sulphorhodamine B
Franke C, et al.
Water Research, 31(10), 2633-2637 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service