Skip to Content
Merck
All Photos(1)

Documents

36154

Supelco

Metamitron

PESTANAL®, analytical standard

Synonym(s):

4-Amino-4,5-dihydro-3-methyl-6-phenyl-1,2,4-triazin-5-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N4O
CAS Number:
Molecular Weight:
202.21
Beilstein:
613129
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water (Karl Fischer)

mp

165-170 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

CC1=NN=C(c2ccccc2)C(=O)N1N

InChI

1S/C10H10N4O/c1-7-12-13-9(10(15)14(7)11)8-5-3-2-4-6-8/h2-6H,11H2,1H3

InChI key

VHCNQEUWZYOAEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metamitron is a 1,2,4-triazine herbicide, which is yellow crystalline in nature and can easily degrade in soil to form stable metabolites. It can be prepared via reaction of N-acetyl-benzylhydrazone with hydroxylamine in the presence of potassium hydroxide in pyridine.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The Pesticide Encyclopedia (2014)
Adsorption of 1, 2, 4-triazine pesticides metamitron and metribuzin on lignin
Ludvik J and Zuman P
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 64, 15-20 (2000)
Jirí Ludvík et al.
Acta crystallographica. Section C, Crystal structure communications, 63(Pt 4), o259-o262 (2007-04-07)
The title structures, both C(10)H(10)N(4)O, are substitutional isomers. The N-N bond lengths are longer and the C=N bond lengths are shorter by ca 0.025 A than the respective average values in the C=N-N=C group of asymmetric triazines; the assessed respective
E Mechant et al.
Communications in agricultural and applied biological sciences, 75(2), 53-59 (2010-01-01)
Chenopodium album L. (fat-hen) with a Ser264-Gly mutation is resistant to photosystem II-inhibiting herbicides like the triazinone metamitron, a key herbicide in sugar beet. In recent years, this resistant biotype may cause unsatisfactory weed control in Belgian sugar beet. However
E Mechant et al.
Communications in agricultural and applied biological sciences, 73(4), 913-917 (2009-02-21)
Chenopodium album L. (fat-hen), a highly competitive and very prolific species, is a common weed in most spring- and summer-sown crops such as maize, sugar beet and vegetables. In the late seventies, C. album stepped into the limelight as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service