Skip to Content
Merck
All Photos(1)

Documents

H43415

Sigma-Aldrich

4-Hydroxy-6-methyl-2-pyrone

98%

Synonym(s):

3,5-Dihydroxysorbic acid δ-lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
Beilstein:
113815
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

188-190 °C (dec.) (lit.)

SMILES string

CC1=CC(O)=CC(=O)O1

InChI

1S/C6H6O3/c1-4-2-5(7)3-6(8)9-4/h2-3,7H,1H3

InChI key

NSYSSMYQPLSPOD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irreversible inactivation of chicken liver fatty acid synthetase by its substrates acetyl and malonyl CoA. Effect of temperature and NADP n fatty acid and triacetic acid lactone synthesis.
K R Srinivasan et al.
Biochemical and biophysical research communications, 99(3), 920-927 (1981-04-15)
J B Spencer et al.
The Biochemical journal, 288 ( Pt 3), 839-846 (1992-12-15)
6-Methylsalicylic acid synthase has been isolated in homogeneous form from Penicillium patulum grown in liquid culture from a spore inoculum. The enzyme is highly susceptible to proteolytic degradation in vivo and in vitro, but may be stabilized during purification by
Wenjuan Zha et al.
Journal of the American Chemical Society, 126(14), 4534-4535 (2004-04-09)
Metabolic pathway engineering is a powerful tool to synthesize structurally diverse and complex chemicals via genetic manipulation of multistep catalytic systems involved in cell metabolism. Here, we report the rational design of a fatty acid biosynthetic pathway, Brevibacterium ammoniagenes fatty
Synthesis of acetoacetyl-CoA by bovine mammary fatty acid synthase.
S Ghayourmanesh et al.
FEBS letters, 132(2), 231-234 (1981-09-28)
Dongming Xie et al.
Biotechnology and bioengineering, 93(4), 727-736 (2005-10-26)
Native g2ps1-encoded 2-pyrone synthase (2-PS) from Gerbera hybrida, a mutant Brevibacterium ammoniagenes fatty acid synthase B (FAS-B) and two different mutants of Penicillium patulum 6-methylsalycilic acid synthase (6-MSAS) are examined to identify the best enzyme to recruit for the microbial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service