Skip to Content
Merck
All Photos(1)

Key Documents

TDF100A

Supelco

Total Dietary Fiber Assay Kit

sufficient for ~100 assays

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12164500
NACRES:
NA.84

usage

sufficient for ~100 assays

Quality Level

technique(s)

photometry: suitable

application(s)

food and beverages
general analytical

storage temp.

2-8°C

Application

This assay determines the total dietary fiber content of food using a combination of enzymatic and gravimetric methods. Samples of dried, fat-free foods are gelatinized with heat stable α-amylase and then enzymatically digested with protease and amyloglucosidase to remove the protein and starch present in the sample. Ethanol is added to precipitate the soluble dietary fiber. The residue is then filtered and washed with ethanol and acetone. After drying, the residue is weighed. Half of the samples are analyzed for protein and the others are ashed. Total dietary fiber is the weight of the residue less the weight of the protein and ash.

Other Notes

Reagents for the determination of total dietary fiber.

Principle

Uses a combination of enzymatic and gravimetric methods. This procedure is based on the method published in Official Methods of Analysis, 15th ed., AOAC, Arlington, VA, Vol. II, Sec. 985.29, 1105 (1990).

Kit Components Only

Product No.
Description

  • α-Amylase, Heat-stable 10 mL

  • Amyloglucosidase 10 mL

  • Celite 50 g

  • Protease 500 mg

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

José Arias-Rico et al.
Foods (Basel, Switzerland), 9(4) (2020-04-05)
The objective of this study is to evaluate the nutritional composition, antioxidant properties, and functional characteristics of two cultivars of xoconostle Opuntia xoconostle F.A.C. Weber in Diguet cv. Cuaresmeño (XC) and Opuntia matudae Scheinvar cv. Rosa (XR). The samples were
Gonul Silav-Tuzlu et al.
Food technology and biotechnology, 59(4), 463-474 (2022-02-10)
In spite of being a significantly growing segment, there are still problems regarding the nutritional, technological and sensory profiles of gluten-free products. Thus, the combination of a variety of functional ingredients is required in order to achieve the desired product
Ashrafi Hossain et al.
Food chemistry, 199, 648-652 (2016-01-19)
This article decribes the nutrient composition of four strawberry genotypes cultivated at the Sher-e-Bangla Agriculture University horticulture farm in Dhaka (Bangladesh). AOAC and standard validated methods were employed to analyse the nutrient composition. Protein, fat and ash contents were found
Isabella Corsato Alvarenga et al.
PloS one, 13(10), e0206090-e0206090 (2018-10-27)
Sorghum is an abundant starch source that has many potential health benefits. Some pet food companies have adopted whole sorghum in their formulations, however sorghum flour and (or) its phenolic rich seed coat might provide added benefit to companion animal
Isabella Corsato Alvarenga et al.
Journal of animal science, 98(4) (2020-03-27)
The growing pet food market is continuously in search for novel ingredients. Legumes such as faba beans (FB) are increasing in popularity and are known to benefit human health, but little is known about their use in pet foods. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service