Skip to Content
Merck
All Photos(1)

Key Documents

45751

Supelco

Diflufenican

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H11F5N2O2
CAS Number:
Molecular Weight:
394.29
Beilstein:
4212494
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Fc1ccc(NC(=O)c2cccnc2Oc3cccc(c3)C(F)(F)F)c(F)c1

InChI

1S/C19H11F5N2O2/c20-12-6-7-16(15(21)10-12)26-17(27)14-5-2-8-25-18(14)28-13-4-1-3-11(9-13)19(22,23)24/h1-10H,(H,26,27)

InChI key

WYEHFWKAOXOVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Diflufenican is a pre- and early post-emergence herbicide used to control dicotyledonous weeds in winter cereals, including Galium aparine, Veronica hederifolia, Veronica persica and Viola arvensis, which are resistant to substituted urea herbicides.

Application

Diflufenican may be used as a reference standard in the determination of diflufenican in water samples using solid-phase microextraction followed by gas chromatography coupled with ion trap-tandem mass spectrometry (SPME-GC-ITD-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Takahiro Yamagishi et al.
Ecotoxicology (London, England), 25(10), 1751-1758 (2016-09-17)
The use of delayed fluorescence intensity as an endpoint for rapid estimation of the effective concentration (EC
Cuticular penetration of foliar-applied diflufenican in Galium aparine L.
Knight H and Kirkwood CR
Pestic. Sci., 33(3), 305-317 (1991)
The effect of diflufenican on lipid metabolism in plants.
I P Ashton et al.
Biochemical Society transactions, 19(3), 320S-320S (1991-08-01)
Pieter Spanoghe et al.
Pest management science, 61(8), 793-798 (2005-05-10)
Herbicides are still used to control weeds on hard surfaces, including municipal, private and industrial sites. Used under unfavourable conditions, especially when rain occurs shortly after application, herbicides may run off to surface waters. Such losses of herbicides from hard
Umi S Ramli et al.
The New phytologist, 184(2), 330-339 (2009-08-07)
* Oil crops are a very important commodity. Although many genes and enzymes involved in lipid accumulation have been identified, much less is known of regulation of the overall process. To address the latter we have applied metabolic control analysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service