Skip to Content
Merck
All Photos(1)

Documents

33379

Supelco

Acetochlor

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-N-ethoxymethyl-N-(2′-ethyl-6′-methylphenyl) acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20ClNO2
CAS Number:
Molecular Weight:
269.77
Beilstein:
2859702
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOCN(C(=O)CCl)c1c(C)cccc1CC

InChI

1S/C14H20ClNO2/c1-4-12-8-6-7-11(3)14(12)16(10-18-5-2)13(17)9-15/h6-8H,4-5,9-10H2,1-3H3

InChI key

VTNQPKFIQCLBDU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetochlor is a chloroacetanilide herbicide effective against soil bacteria. This selective pre-emergence herbicide inhibits the elongation of C18 and C16 fatty acids species to form very long-chain fatty acids (VLCFAs). It is commonly used for weed control management and yellow nutsedge in corn plantation.

Application

Acetochlor may be used as a reference standard to deternine acetochlor in water and soil samples using high-performance liquid chromatography and reversed-phase liquid chromatography coupled with electrospray ionization mass spectrometry, respectivley.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Kidney, Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wei Ouyang et al.
Environment international, 130, 104858-104858 (2019-06-19)
In several watersheds, agricultural activities are the cause of pollution, mainly due to the discharge of herbicides. Often, these herbicide plumes are transported to the surrounding bays. Samples of water, suspended particulate sediments (SPSs), and sediments from 37 sites in
Analysis and detection of the new corn herbicide acetochlor in river water and rain.
Capel P D, et al.
Environmental Science & Technology, 29(6), 1702-1705 (1995)
Jinhong Lü et al.
Journal of agricultural and food chemistry, 60(26), 6463-6470 (2012-06-13)
The sustained release and reduced leaching of herbicides is expected for enhancing their efficacy and minimizing their pollution. For this purpose, the rice straw biochar made at a relatively low temperature (350 °C) (RS350) was used simultaneously as the carrier
Francisco Bedmar et al.
Environmental toxicology and chemistry, 30(9), 1990-1996 (2011-06-22)
Understanding herbicide sorption within soil profiles is the first step to predicting their behavior and leaching potential. Laboratory studies were conducted to determine the influence of surface and subsurface soil properties on acetochlor, atrazine, and S-metolachlor sorption. Soil samples were
Hugo Ochoa-Acuña et al.
Environmental health perspectives, 117(10), 1619-1624 (2009-12-19)
Atrazine and other corn herbicides are routinely detected in drinking water. Two studies on potential association of atrazine with small-for-gestational-age (SGA) and preterm birth prevalence found inconsistent results. Moreover, these studies did not control for individual-level potential confounders. Our retrospective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service