Skip to Content
Merck
All Photos(1)

Key Documents

36165

Supelco

Metribuzin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N4OS
CAS Number:
Molecular Weight:
214.29
Beilstein:
746650
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

Light sensitive

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

storage condition

under inert gas (Argon)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CSC1=NN=C(C(=O)N1N)C(C)(C)C

InChI

1S/C8H14N4OS/c1-8(2,3)5-6(13)12(9)7(14-4)11-10-5/h9H2,1-4H3

InChI key

FOXFZRUHNHCZPX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Metribuzin is a selective triazine herbicide, which can be used both as a preemergence and postemergence herbicide. It can generally find applications in controlling broadleaved weeds and annual grasses in cultivation of crops like sugarcane, wheat and soyabean. Its mode of action involves the inhibition of photosynthesis process in plants.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Effect of soil amendments on sorption and mobility of metribuzin in soils
Majumdar K and Singh N
Chemosphere, 66(4), 630-637 (2007)
Effect of metribuzin, butachlor and chlorimuron-ethyl on amino acid and protein formation in wheat and maize seedlings
Alla N.M.M, et al.
Pesticide Biochemistry and Physiology, 90(1), 8-18 (2008)
Annette E Rosenbom et al.
Environmental science & technology, 43(9), 3221-3226 (2009-06-19)
In a regulatory context, numerical models are increasingly employed to quantify leaching of pesticides and their metabolites. Although the ability of these models to accurately simulate leaching of pesticides has been evaluated, little is known about their ability to accurately
Celia Maqueda et al.
Journal of agricultural and food chemistry, 56(24), 11941-11946 (2008-12-05)
Metribuzin is an herbicide widely used for weed control that has been identified as a groundwater pollutant. It contaminates the environment even when it is used according to the manufacturer's instructions. To reduce herbicide leaching and increase weed control, new
Celia Maqueda et al.
Journal of agricultural and food chemistry, 57(8), 3273-3278 (2009-04-17)
Metribuzin (MTB) is a herbicide widely used for weed control in growing soybeans and other crops and has been identified in many parts of the world as a groundwater contaminant. To prepare controlled-release formulations (CRFs) of MTB, it was entrapped

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service