Skip to Content
Merck
All Photos(1)

Documents

34210

Supelco

Nicosulfuron

PESTANAL®, analytical standard

Synonym(s):

1-(4,6-Dimethoxy-2-pyrimidinyl)-3-[3-(dimethylcarbamoyl)-2-pyridylsulfonyl]urea, 2-[(4,6-Dimethoxypyrimidin-2-ylcarbamoyl)sulfamoyl]-N,N-dimethylnicotinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N6O6S
CAS Number:
Molecular Weight:
410.41
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cc(OC)nc(NC(=O)NS(=O)(=O)c2ncccc2C(=O)N(C)C)n1

InChI

1S/C15H18N6O6S/c1-21(2)13(22)9-6-5-7-16-12(9)28(24,25)20-15(23)19-14-17-10(26-3)8-11(18-14)27-4/h5-8H,1-4H3,(H2,17,18,19,20,23)

InChI key

RTCOGUMHFFWOJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nicosulfuron belongs to the group of sulfonylurea herbicides, which can be largely used to control grasses and broad leaved weed species in crop protection like in corn.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lieqing Yang et al.
Analytica chimica acta, 670(1-2), 72-77 (2010-08-06)
A new molecularly imprinted stir bar was prepared using nicosulfuron, a sulfonylurea herbicide, as a template. To achieve the selective and direct extraction of a target analyte from aqueous samples, several main parameters, including extraction time, pH value and contents
Eunsoo Choe et al.
Pest management science, 76(9), 3012-3019 (2020-04-06)
Nicosulfuron, a sulfonylurea herbicide widely used for grass weed control in corn production, injures some sweet corn hybrids and inbreds. A specific cytochrome P450 (P450), CYP81A9, is suggested to be responsible for sensitivity to nicosulfuron and other P450-metabolized herbicides. Corn
Jussara B Regitano et al.
Journal of agricultural and food chemistry, 56(14), 5801-5805 (2008-06-24)
Sorption-desorption interactions of pesticides with soil determine their availability for transport, plant uptake, and microbial degradation. These interactions are affected by the physical-chemical properties of the pesticide and soil, and for some pesticides, their residence time in the soil. This
Adsorption and desorption of nicosulfuron in soils
Gonzalez.MJ and Ukrainczyk L
Journal of Environmental Quality, 25, 1186-1192 (1996)
P Saglio et al.
Archives of environmental contamination and toxicology, 41(2), 192-200 (2001-07-20)
The immediate behavioral responses of goldfish (Carassius auratus) to pesticide-contaminated flows were recorded in a countercurrent olfactometer. In addition, electro-olfactograms were recorded from the epithelial surface of the olfactory rosette as a preliminary check for the olfactory sensitivity of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service