Skip to Content
Merck
All Photos(1)

Documents

45330

Supelco

Atrazine

PESTANAL®, analytical standard

Synonym(s):

2-Chloro-4-ethylamino-6-isopropylamino-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14ClN5
CAS Number:
Molecular Weight:
215.68
Beilstein:
612020
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

solid phase extraction (SPE): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(Cl)nc(NC(C)C)n1

InChI

1S/C8H14ClN5/c1-4-10-7-12-6(9)13-8(14-7)11-5(2)3/h5H,4H2,1-3H3,(H2,10,11,12,13,14)

InChI key

MXWJVTOOROXGIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dmitry P Bazhanov et al.
BMC microbiology, 16(1), 265-265 (2016-11-09)
Soil populations of bacteria rapidly degrading atrazine are critical to the environmental fate of the herbicide. An enrichment bias from the routine isolation procedure prevents studying the diversity of atrazine degraders. In the present work, we analyzed the occurrence, diversity
Dmitry P Bazhanov et al.
Applied microbiology and biotechnology, 101(17), 6809-6820 (2017-07-13)
Our previous research found that culturable atrazine degraders associated with maize roots were dominated by genetically similar strains of Arthrobacter ureafaciens, suggesting their rhizosphere competence. The present study aimed to assess the root-colonizing capacity of strain A. ureafaciens DnL1-1 and
Laura Lui de Andrade et al.
The Science of the total environment, 663, 548-559 (2019-02-06)
Atrazine (ATZ) is a widely used herbicide that has the potential to contaminate the environment and cause deleterious effects on non-target organisms. Release systems for ATZ have been developed to minimize this contamination, such as nanocapsules prepared with poly (ε-caprolactone)
Sarah R Marcus et al.
Journal of insect physiology, 91-92, 18-25 (2016-06-19)
Atrazine is the one of the most widely used herbicides in the United States and non-target organisms may encounter it in the environment. Atrazine is known to affect male reproduction in both vertebrates and invertebrates but less is known about
Andrea Vogel et al.
Journal of insect physiology, 72, 14-21 (2014-12-03)
Atrazine is a commonly utilized herbicide to control broadleaf weeds in the agricultural setting. It can, however, have negative effects on male reproductive performance in a variety of vertebrate species. Much less is known, however, about the effects of atrazine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service