Skip to Content
Merck
All Photos(1)

Documents

880129C

Avanti

DSPE-PEG(2000) Biotin

Avanti Research - A Croda Brand 880129C

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[biotinyl(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

liquid

packaging

pkg of 1 × 1 mL (880129C-10mg)
pkg of 1 × 2 mL (880129C-50mg)
pkg of 1 × 2.5 mL (880129C-25mg)
pkg of 1 × 4 mL (880129C-100mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880129C

concentration

10 mg/mL (880129C-10mg)
10 mg/mL (880129C-25mg)
25 mg/mL (880129C-100mg)
25 mg/mL (880129C-50mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOOO/C=C\NC(CCCC[C@](SC[C@@]1(NC(N2)=O)[H])([C@]12[H])[H])=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=C.C=

General description

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[biotinyl(polyethylene glycol)-2000] (DSPE-PEG(2000) based micelles display a spheroids shape and are effective agents for drug delivery. Their size ranges between 18 nm to 90 nm. It is used with functional groups like biotin added as conjugate.

Application

DSPE-PEG(2000) Biotin has been used in the generation of biotinylated lipid-coated microbubbles for adhering to human umbilical vein endothelial cells (HUVECs) and chicken embryo for ultrasound studies. It has also been used for the generation of giant unilamellar lipid vesicles (GUVs) for live cell fusion studies.

Biochem/physiol Actions

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[biotinyl(polyethylene glycol)-2000] (DSPE-PEG(2000)) Biotin is used in capsule surface modification.

Packaging

5 mL Clear Glass Sealed Ampule (880129C-100mg)
5 mL Clear Glass Sealed Ampule (880129C-10mg)
5 mL Clear Glass Sealed Ampule (880129C-25mg)
5 mL Clear Glass Sealed Ampule (880129C-50mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SPIO labeling of endothelial cells using ultrasound and targeted microbubbles at diagnostic pressures
Skachkov I, et al.
Testing, 13(9), e0204354-e0204354 (2018)
In vivo characterization of ultrasound contrast agents: microbubble spectroscopy in a chicken embryo
Faez T, et al.
Ultrasound in Medicine & Biology, 38(9), 1608-1617 (2012)
Hot-nanoparticle-mediated fusion of selected cells
Bahadori A, et al.
Nano Research, 10(6), 2034-2045 (2017)
Wen-Qun Li et al.
International journal of nanomedicine, 14, 8345-8360 (2019-11-07)
The protective role of puerarin (PUE) against myocardial infarction is closely related to its regulation on mitochondria. However, free PUE can hardly reach the mitochondria of ischemic cardiomyocytes due to the lack of mitochondrial targeting of PUE. Here PUE was
Bolin Wu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(12), 14129-14136 (2019-10-29)
The aim of this study was to establish a novel targeting nanobubble (TNB) conjugated with small interfering RNA (siRNA)-cyanine 5 (Cy5) and to validate its theranostic ability in vivo. The TNB conjugated with neuroepithelial transforming gene 1 (NET-1) siRNA-Cy5 was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service