GF57898760
Zinc
wire reel, 0.5m, diameter 1.0mm, 99.999%
Synonym(s):
Zinc, ZN005133
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
vapor pressure
1 mmHg ( 487 °C)
Assay
99.999%
form
wire
manufacturer/tradename
Goodfellow 578-987-60
resistivity
5.8 μΩ-cm, 20°C
L × diam.
0.5 m × 1 mm
bp
907 °C (lit.)
mp
420 °C (lit.)
density
7.133 g/mL at 25 °C (lit.)
SMILES string
[Zn]
InChI
1S/Zn
InChI key
HCHKCACWOHOZIP-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Douglas J Lanska et al.
Neurology, 82(13), 1175-1179 (2014-04-02)
Zinc-induced myeloneuropathy was recently (re)discovered and its pathophysiology elaborated as resulting from secondary copper deficiency. However, myelopathy was a recognized problem among European zinc-smelter workers in the late 19th century, although these early reports have been overlooked in recent studies
J Granell
The Journal of sports medicine and physical fitness, 54(2), 232-237 (2014-02-11)
The role of zinc and copper has been shown essential in the scope of physical exercise. The outcomes of the studies about changes in the concentrations of these elements in blood, in physical effort situations, are sometimes discordant and seem
M Munirul Islam et al.
The Journal of nutrition, 144(5), 771-776 (2014-03-07)
Information is needed on breast milk zinc concentration and amount of zinc transferred to the infant in relation to the time since birth for both appropriate-for-gestational-age (AGA) and small-for-gestational-age (SGA) infants. Breast milk zinc concentration and total milk volume were
Shinya Tashiro et al.
Biochemistry, 53(14), 2218-2220 (2014-04-05)
Mutations of DJ-1 cause familial Parkinson's disease (PD), although the role of DJ-1 in PD remains unresolved. Very recent reports have shown that DJ-1 interacts with copper ions. This evidence opens new avenues to understanding the function of DJ-1 and
Venus Singh Mithu et al.
The Journal of biological chemistry, 289(16), 11122-11131 (2014-03-07)
Amyloid β (Aβ) fibrillar deposits in the brain are a hallmark of Alzheimer disease (AD). Curcumin, a common ingredient of Asian spices, is known to disrupt Aβ fibril formation and to reduce AD pathology in mouse models. Understanding the structural
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service