Skip to Content
Merck
All Photos(1)

Key Documents

356018

Sigma-Aldrich

Zinc

foil, thickness 0.25 mm, 99.9% trace metals basis

Synonym(s):

Zn

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 487 °C)

Quality Level

Assay

99.9% trace metals basis

form

foil

reaction suitability

core: zinc
reagent type: catalyst

resistivity

5.8 μΩ-cm, 20°C

thickness

0.25 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Application

Zinc can be used as a catalyst:
  • For the reduction of nitrobenzenes to aniline derivatives in the presence of ammonium chloride[1].
  • To synthesize β-hydroxy esters using ketones and α-bromoacetates via Reformatsky reaction[2].
  • For the transesterification of ethyl acetate and benzyl alcohol to synthesize benzyl acetate in the presence of imidazole[3].

Quantity

  • 50 × 50 mm (approximately 4.5 g)
  • 100 × 100 mm (approximately 18 g)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and convenient conversion of nitroarenes to anilines under microwave conditions using nonprecious metals in mildly acidic medium
Keenan C, et al.
Synthetic Communications, 47, 1085-1089 (2017)
Transesterification reactions catalyzed by a recyclable heterogeneous zinc/imidazole catalyst
Nakatake D, et al.
advanced synthesis and catalysis, 358, 2569-2574 (2016)
Shigeki Seto et al.
Bioorganic & medicinal chemistry, 20(3), 1188-1200 (2012-01-21)
The design, synthesis, and evaluation of 6-6-7 tricyclic quinolones containing the strained spirocycle moiety aiming at the GSK-3β inhibitor were described. Among the synthesized compounds, 44, having a cyclobutane ring on a spirocycle, showed excellent GSK-3β inhibitory activity in both
Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
M E Kuyumcu et al.
European review for medical and pharmacological sciences, 17(11), 1467-1471 (2013-06-19)
Zinc is one of the most important elements for human body. Zinc deficiency can occur in any age, if it is seen in elderly its clinical results can be more harmful due to already diminished functions. Some studies showed zinc

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service