GF52959785
Zinc
foil, 300x300mm, thickness 0.1mm, as rolled, 99.95+%
Synonym(s):
Zinc, ZN000250
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
vapor pressure
1 mmHg ( 487 °C)
Assay
99.95%
form
foil
manufacturer/tradename
Goodfellow 529-597-85
resistivity
5.8 μΩ-cm, 20°C
size × thickness
300x300 mm × 0.1 mm
bp
907 °C (lit.)
mp
420 °C (lit.)
density
7.133 g/mL at 25 °C (lit.)
SMILES string
[Zn]
InChI
1S/Zn
InChI key
HCHKCACWOHOZIP-UHFFFAOYSA-N
General description
For updated SDS information please visit www.goodfellow.com.
Legal Information
Product of Goodfellow
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chenglin Yan et al.
The journal of physical chemistry. B, 110(51), 25850-25855 (2006-12-22)
Vertically aligned ZnO nanorods with uniform diameter and length have been synthesized on a zinc foil substrate with ammonium persulfate as oxidant via a facile, larger scale production and inexpensively synthesized method without any templates or additives. SEM and XRD
S Rondot et al.
Science (New York, N.Y.), 263(5154), 1739-1741 (1994-03-25)
An x-ray projection microscope equipped with a charge-coupled device camera allows direct observation of zinc (Zn(2+)) ions diffusing in aqueous hydrochloric acid solution during the corrosion of zinc foil and pellets. Time series of microradiographic images with a lateral resolution
Tandra Ghoshal et al.
Journal of nanoscience and nanotechnology, 9(9), 5586-5591 (2009-11-26)
Nanowire arrays of zinc oxide were synthesized on zinc foil by a simple thermal evaporation process. Morphologies and sizes of the synthesized nanostructures were varied by varying the reaction time and the surface roughness of the substrate. Self-catalytic liquid-solid mechanism
A new vapor-phase hydrothermal method to concurrently grow ZnO nanotube and nanorod array films on different sides of a zinc foil substrate.
Haimin Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(17), 5165-5169 (2012-03-21)
Xufeng Wu et al.
Chemical communications (Cambridge, England), (15)(15), 1655-1657 (2006-04-04)
Highly oriented ZnO nanoneedle/nanorods arrays have been fabricated by direct oxidation of zinc foil in alkaline zincate ion solution at near room temperature (20 degrees C for nanoneedles, 30 degrees C for nanorods).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service