Skip to Content
Merck
All Photos(3)

Key Documents

267635

Sigma-Aldrich

Zinc

shot, 5 mm, 99.999% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

1 mmHg ( 487 °C)

Assay

99.999% trace metals basis

form

shot

resistivity

5.8 μΩ-cm, 20°C

particle size

5 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sara Eyal et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 50(5), 798-806 (2009-05-01)
Studies in rodents indicate that the disruption of P-glycoprotein (P-gp) function increases drug distribution into the developing fetus and organs such as the brain. To simultaneously and serially evaluate the effect of P-gp activity and inhibition on the tissue distribution
Casper Groth et al.
Development (Cambridge, England), 140(14), 3018-3027 (2013-06-21)
Developmental patterning requires the precise interplay of numerous intercellular signaling pathways to ensure that cells are properly specified during tissue formation and organogenesis. The spatiotemporal function of the Notch signaling pathway is strongly influenced by the biosynthesis and intracellular trafficking
I A Popov et al.
Molekuliarnaia biologiia, 47(3), 498-504 (2013-07-31)
Alzheimer's disease, is a lethal neurodegenerative pathology, characterized by the formation of soluble neurotoxic oligomers of the human amyloid-beta peptide Abeta which get accumulated forming polymeric extracellular aggregates (so-called amyloid plaques). The isomerized at aspartate 7 isoform of the human
Cigdem Karakukcu et al.
Clinical laboratory, 59(5-6), 557-562 (2013-07-20)
The aim of this study was to assess the acute and chronic effects of vigorous physical activity, with and without zinc supplementation, on distribution of elements in young amateur boxers. The 8 week intervention trial experiment was designed with 32
Nalini Pandey et al.
Indian journal of experimental biology, 51(7), 548-555 (2013-08-01)
Black gram plants subjected to varying levels of Zn supply (0.01 to 10 microM Zn) showed optimum growth and dry matter yield in plants receiving 1 microM Zn. The dry matter yield of plants decreased in plants receiving 0.01 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service