Skip to Content
Merck
All Photos(3)

Key Documents

243469

Sigma-Aldrich

Zinc

granular, 20-30 mesh, ACS reagent, ≥99.8%

Synonym(s):

Zn

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

ACS reagent

Quality Level

vapor pressure

1 mmHg ( 487 °C)

Assay

≥99.8%

form

granular

reaction suitability

core: zinc
reagent type: catalyst

resistivity

5.8 μΩ-cm, 20°C

particle size

20-30 mesh

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

cation traces

Fe: ≤0.01%
Pb: ≤0.01%

suitability

passes application test for determination of arsenic (As)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

General description

Zinc granules are pure metallic zinc in a granular form, they provide a high surface area for increased reactivity. They serve as a reducing agent, catalyst, or precursor for various chemical reactions. Zinc granules are commonly used for synthesis of organometallic compounds. It reacts with organic halides to form alkyl or aryl zinc compounds, which serve as powerful nucleophiles in carbon-carbon bond formations. This reaction, known as the Barbier reaction, enables the introduction of diverse functional groups into organic molecules.Zinc granules also find application in the synthesis of pharmaceuticals and fine chemicals. They can be employed in the reduction of functional groups like nitro, nitrile, or azide to their corresponding amines. Additionally, zinc granules are involved in other carbon-carbon bond forming reactions, such as the Reformatsky reaction and the Simmons-Smith reaction.

Application

Zinc granules can be used to prepare:

  • organozinc reagents commonly used in Negishi or Fukuyama couplings
  • Reformatsky reagents
  • Simmons-Smith reagents

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Advanced Organic Chemistry: Reactions, Mechanisms, and Structure
March, J
Advanced Organic Chemistry (2007)
Use of activation methods for organozinc reagents.
Erdik E
Tetrahedron, 43(10), 2203-2212 (1987)
Reagents for Organic Synthesis
L. F. Fieser and M. Fieser
Reagents for Organic Synthesis, 1 (1967)
A scalable zinc activation procedure using dibal-h in a reformatsky reaction.
Girgis M J
Organic Process Research & Development, 13(6), 1094-1099 (2009)
A Convenient Procedure for the Preparation of Reactive Zinc for the Reformatsky Reaction.
Bouhel E
Synthetic Communications, 21(1), 133-136 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service