Skip to Content
Merck
All Photos(1)

Key Documents

64430

Sigma-Aldrich

DL-Methionine sulfoxide

≥98.5% (NT)

Synonym(s):

dl-Methionine sulfoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SOCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
165.21
Beilstein:
1723793
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.5% (NT)

reaction suitability

reaction type: solution phase peptide synthesis

mp

~240 °C (dec.)

application(s)

peptide synthesis

SMILES string

CS(=O)CCC(N)C(O)=O

InChI

1S/C5H11NO3S/c1-10(9)3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

InChI key

QEFRNWWLZKMPFJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-Methionine sulfoxide is an oxidation product of DL-methionine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The replaceability of dl-methionine in the diet of the albino rat with dl-methionine sulfone and dl-methionine methylsulfonium chloride.
Bennet AM, et al.
The Journal of Biological Chemistry, 141(4), 573-578 (1941)
Bart Ghesquière et al.
Molecular & cellular proteomics : MCP, 10(5), M110-M110 (2011-03-17)
We here present a new method to measure the degree of protein-bound methionine sulfoxide formation at a proteome-wide scale. In human Jurkat cells that were stressed with hydrogen peroxide, over 2000 oxidation-sensitive methionines in more than 1600 different proteins were
Jérémy Couturier et al.
FEBS letters, 586(21), 3894-3899 (2012-10-02)
A natural fusion occurring between two tandemly repeated glutaredoxin (Grx) modules and a methionine sulfoxide reductase A (MsrA) has been detected in Gracilaria gracilis. Using an in vivo yeast complementation assay and in vitro activity measurements, we demonstrated that this
Jackob Moskovitz et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 161(4), 406-414 (2012-01-11)
The blind subterranean mole rat (Spalax ehrenbergi) exhibits a relatively long life span, which is attributed to an efficient antioxidant defense affording protection against accumulation of oxidative modifications of proteins. Methionine residues can be oxidized to methionine sulfoxide (MetO) and
Fiona M Sansom et al.
PloS one, 8(2), e56064-e56064 (2013-02-26)
Leishmania are protozoan parasites that proliferate within the phagolysome of mammalian macrophages. While a number of anti-oxidant systems in these parasites have been shown to protect against endogenous as well as host-generated reactive oxygen species, the potential role of enzymes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service