M9375
D-Methionine
≥98% (HPLC)
Synonym(s):
(R)-2-Amino-4-(methylmercapto)butyric acid, D-2-Amino-4-(methylthio)butanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C5H11NO2S
CAS Number:
Molecular Weight:
149.21
Beilstein:
1722293
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
powder
color
white
mp
273 °C
application(s)
cell analysis
peptide synthesis
SMILES string
CSCC[C@@H](N)C(O)=O
InChI
1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m1/s1
InChI key
FFEARJCKVFRZRR-SCSAIBSYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
D-methionine has been shown to rescue noise-induced hearing loss.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Eleni Ntone et al.
Journal of colloid and interface science, 583, 459-469 (2020-10-05)
Plants offer a vast variety of protein extracts, typically containing multiple species of proteins that can serve as building blocks of soft materials, like emulsions. However, the role of each protein species concerning the formation of emulsions and interfaces with
Kathleen Campbell et al.
Hearing research, 282(1-2), 138-144 (2011-09-20)
We have previously reported rescue from noise-induced auditory brainstem response (ABR) threshold shifts with d-methionine (d-met) administration 1 h after noise exposure. The present study investigated further d-met rescue intervals at 3, 5 and 7 h post-noise exposure. Chinchillas laniger were exposed
Audrey S Vanhove et al.
Applied and environmental microbiology, 86(11) (2020-03-30)
While only a subset of Vibrio cholerae strains are human diarrheal pathogens, all are aquatic organisms. In this environment, they often persist in close association with arthropods. In the intestinal lumen of the model arthropod Drosophila melanogaster, methionine and methionine
Calvin Wu et al.
Journal of neural transmission (Vienna, Austria : 1996), 121(7), 683-693 (2014-02-18)
Antioxidants are well known for their neuroprotective properties against reactive oxygen species in cortical neurons and auditory cells. We recently identified L-carnitine and D-methionine to be among agents that provide such protection. Here, we investigated their neuronal modulatory actions. We
Yi-Sin Wong et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 32(6), e13803-e13803 (2020-01-29)
Cisplatin is a widely used antineoplastic drug. However, cisplatin-induced dyspepsia syndromes, including delayed gastric emptying, gastric distension, early satiety, nausea, and vomiting, often force patients to take doses lower than those prescribed or even refuse treatment. D-methionine has an appetite-enhancing
Chromatograms
application for HPLCapplication for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service