Skip to Content
Merck
All Photos(1)

Key Documents

M0876

Sigma-Aldrich

L-Methionine sulfone

≥98% (TLC)

Synonym(s):

L-2-Amino-4-(methylsulfonyl)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO4S
CAS Number:
Molecular Weight:
181.21
Beilstein:
1725510
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

L-Methionine sulfone,

Assay

≥98% (TLC)

form

powder or crystals

mp

275 °C

application(s)

detection
peptide synthesis

SMILES string

CS(=O)(=O)CC[C@H](N)C(O)=O

InChI

1S/C5H11NO4S/c1-11(9,10)3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1

InChI key

UCUNFLYVYCGDHP-BYPYZUCNSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

L-Methionine sulfone may be used to study pupae development of the silkworm Bombyx mori L. L-Methionine sulfone may be used as a complexing agent to study conformational structures and stereospecificity of enzymes such as glutamate synthase and gamma-glutamyl transpeptidase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuri V Bobryshev et al.
Atherosclerosis, 215(2), 355-365 (2011-02-08)
The origin of smooth muscle cells in developing atherosclerotic lesions is a controversial topic with accumulating evidence indicating that at least some arterial smooth muscle cells might originate from bone marrow-derived smooth muscle cell precursors circulating in the blood. The
Rasmus Dahl-Lassen et al.
Plant methods, 14, 8-8 (2018-01-30)
The amino acid profile of plants is an important parameter in assessments of their growth potential, resource-use efficiency and/or quality as food and feed. Screening studies may involve large number of samples but the classical amino acid analysis is limited
Binning Wu et al.
Frontiers in plant science, 11, 619598-619598 (2021-02-16)
Increasing incidence of chronic diseases in the 21st century has emphasized the importance of developing crops with enhanced nutritional value. Plant-based diets are associated with reduced incidence of many chronic diseases. The growing population and increased food demand have prioritized
M Sta et al.
Neurobiology of disease, 42(3), 211-220 (2011-01-12)
Increasing evidence suggests a role for the immune system in amyotrophic lateral sclerosis (ALS). To determine the extent of the immune activation in ALS we analyzed the expression and cellular distribution of components of innate and adaptive immunity in spinal
Lake-Ee Quek et al.
iScience, 23(2), 100855-100855 (2020-02-15)
Cellular metabolism is dynamic, but quantifying non-steady metabolic fluxes by stable isotope tracers presents unique computational challenges. Here, we developed an efficient 13C-tracer dynamic metabolic flux analysis (13C-DMFA) framework for modeling central carbon fluxes that vary over time. We used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service