Skip to Content
Merck
All Photos(3)

Key Documents

40234

Sigma-Aldrich

N,N-Dimethyldodecylamine N-oxide

BioXtra, ≥99.0% (NT)

Synonym(s):

DDAO, LDAO, Lauryldimethylamine N-oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11N(O)(CH3)2
CAS Number:
Molecular Weight:
229.40
Beilstein:
1769927
EC Number:
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.26

description

non-ionic

Quality Level

product line

BioXtra

Assay

≥99.0% (NT)

form

powder

mol wt

229.40 g/mol

technique(s)

protein quantification: suitable

impurities

≤0.03% peroxides (as H2O2)

CMC

1-2 mM

mp

132-133 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤100 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCC[N+](C)(C)[O-]

InChI

1S/C14H31NO/c1-4-5-6-7-8-9-10-11-12-13-14-15(2,3)16/h4-14H2,1-3H3

InChI key

SYELZBGXAIXKHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

N,N-Dimethyldodecylamine N-oxide is a non-denaturing zwitterionic surfactant with a critical micelle concentration (CMC) of approximately 1 mM. It is used to solubilize proteins and to study the conformation and molecular interactions of macromolecules.

Application

N,N-Dimethyldodecylamine N-oxide has been used in the cell lysis and resuspension of cell pellets for western blot analysis.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Karlovská et al.
Biophysical chemistry, 119(1), 69-77 (2005-10-15)
Sarcoplasmic reticulum Ca-transporting ATPase (EC 3.6.1.38) was isolated from rabbit white muscle, purified and reconstituted into vesicles of synthetic diacylphosphatidylcholines with monounsaturated acyl chains using the cholate dilution method. In fluid bilayers at 37 degrees C, the specific activity of
Rie Kakehashi et al.
Journal of colloid and interface science, 289(2), 498-503 (2005-08-23)
In the hydrogen ion titration of micelles, the degree of ionization of the micelle at a given pH has to be evaluated to obtain a pKa value of micelles (Ka being the proton dissociation constant) at the pH. We compared
Eugenio Daviso et al.
Journal of the American Chemical Society, 133(42), 16754-16757 (2011-10-04)
Several techniques rely on electron-nuclear interactions to boost the polarization of nuclear spins in the solid phase. Averaging out of anisotropic interactions as a result of molecular tumbling strongly reduces the applicability of such hyperpolarization approaches in liquids. Here we
Benesh Joseph et al.
The Journal of biological chemistry, 286(47), 41008-41017 (2011-09-29)
ATP-binding cassette (ABC) transporters are ubiquitous integral membrane proteins that translocate substrates across cell membranes. The alternating access of their transmembrane domains to opposite sides of the membrane powered by the closure and reopening of the nucleotide binding domains is
Stefanie Keis et al.
Journal of bacteriology, 188(11), 3796-3804 (2006-05-19)
The F(1)F(o)-ATP synthases of alkaliphilic bacteria exhibit latent ATPase activity, and for the thermoalkaliphile Bacillus sp. strain TA2.A1, this activity is intrinsic to the F(1) moiety. To study the mechanism of ATPase inhibition, we developed a heterologous expression system in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service