Skip to Content
Merck
All Photos(4)

Documents

M9500

Sigma-Aldrich

DL-Methionine

≥99%, suitable for cell culture and insect cell cutlure

Synonym(s):

(±)-2-Amino-4-(methylmercapto)butyric acid, DL-2-Amino-4-(methylthio)butanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
149.21
Beilstein:
636185
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

DL-Methionine, ≥99%

Assay

≥99%

form

powder

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

color

white

mp

280 °C (dec.) (lit.)

solubility

H2O: soluble

application(s)

peptide synthesis

SMILES string

CSCCC(N)C(O)=O

InChI

1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)

InChI key

FFEARJCKVFRZRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-Methionine is an essential amino acid containing sulphur. Methionine consists of an asymmetric carbon and exists as D (dextrogyre) and L (levogyre) optical isomers. The L-methionine is considered as biologically active. The racemic mixture of D and L-isomers forms DL-methionine, which is the commercially available methionine.

Application

DL-Methionine has been used to study its effect on the growth and feed utilization of Nile tilapia. It has also been used as a part of standard diet in animal models.

Biochem/physiol Actions

DL-Methionine is an amino acid that has a role in the defence of oxidative stress.
Methionine offers protection against disorders related to hair, skin and nails. It elevates lecithin production in the liver and thereby reduces cholesterol level. It naturally serves as a heavy metal chelating agent, regulating ammonia concentration in the urine. This ameliorates bladder irritation. Methionine is also known to induce hair growth.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DL-Methionine from Methional and Hydrogen Cyanide - Cost Analysis - Methionine E11A, 6-6 (2016)
Dietary Crystalline DL-Methionine Supplementation of Oilseed Meal-Based Diets: Effects on Growth and Feed Utilization of Juvenile Nile Tilapia (Oreochromis niloticus L.)
Agbo NW, et al.
American Journal of Experimental Agriculture, 4(11), 1385-1396 (2014)
Gel growth and characterization of beta-DL-methionine
Ramachandran E and Natarajan S
Crystal Research and Technology, 41(4), 411-415 (2006)
Glucose homeostasis in mice is transglutaminase 2 independent
Iismaa SE, et al.
PLoS ONE, 8(5), e63346-e63346 (2013)
Partial replacement of fishmeal and fish oil by algal meals in diets of red drum Sciaenops ocellatus
Perez-Velazquez M, et al.
Aquaculture (Amsterdam, Netherlands), 487, 41-50 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service