Skip to Content
Merck
All Photos(1)

Documents

643203

Sigma-Aldrich

Gold coated glass slide

99.999% (Au), layer thickness 100 Å

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Au
CAS Number:
Molecular Weight:
196.97
EC Number:
MDL number:
UNSPSC Code:
12141717
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% (Ti)
99.999% (Au)

L × W × thickness

3 in. × 1 in. × 0.7 mm

layer thickness

100 Å

transmittance

380-700, >91% (for the glass microscope slide alone, 55-59% @ 500 nm with the Ti/Au coating)
60-64%

refractive index

n20/400 1.52 (lit.)

matrix attachment

Titanium, as adhesion layer used to bind the gold to the aluminosilicate glass cover slip

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Gold coated microscope slide can be used as a substrate material that facilitates the formation of highly active surfaces for surface enhanced Raman scattering (SERS) measurements.
Titanium adhesion layer used to bind the gold to an aluminasilicate glass microscope slide. Titanium thickness ~20 angstroms.

Features and Benefits

The gold surface shows a tendency towards <111> orientation.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Facile preparations of au nanostructures for SERS
Technical Digest of the Eighteenth Micro-optics Conference, 414, 1-2 (2013)
Facile preparation of SERS and catalytically active Au nanostructures using furfuryl derivatives
Kim K, et al.
Applied Surface Science, 414, 325-334 (2017)
Tae-Sik Cho et al.
Journal of nanoscience and nanotechnology, 13(5), 3711-3714 (2013-07-19)
The crystallization of Au/glass ultrathin films for surface plasmon resonance (SPR) biosensor has been studied using synchrotron X-ray scattering and field emission scanning electron microscope. In films thinner than 30 nm, crystallized Au grains with [111] preferred orientation were formed
Midas touch in cardiology.
Marianna Karamanou et al.
European heart journal, 34(20), 1463-1464 (2013-07-11)
Nidhi Gupta et al.
Journal of nanoscience and nanotechnology, 13(7), 4917-4924 (2013-08-02)
The preparation and photocatalytic reductive ability of a new spongy flower or cotton bud like amorphous CdS microstructures (size = 200-300 nm) having intermediate crystal phase between cubic and hexagonal structure and possesses much larger surface area ca. 119 m2

Articles

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.

Protocols

SAMs are versatile surface coatings for chemical resistance, bio-compatibility, sensitization, and molecular recognition for sensors and nano fabrication.

SAMs are versatile surface coatings for chemical resistance, bio-compatibility, sensitization, and molecular recognition for sensors and nano fabrication.

SAMs are versatile surface coatings for chemical resistance, bio-compatibility, sensitization, and molecular recognition for sensors and nano fabrication.

SAMs are versatile surface coatings for chemical resistance, bio-compatibility, sensitization, and molecular recognition for sensors and nano fabrication.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service