Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gold coated glass slide
Linear Formula:
Au
CAS Number:
7440-57-5
Molecular Weight:
196.97
Products
Product Category
slides (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Purity
Length (in)
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
643203
Gold coated glass slide,
99.999% (Au), layer thickness 100 Å
Expand
View Pricing
643246
Gold coated glass slide,
layer thickness 1000 Å, 99.999% (Au)
Expand
View Pricing
900554
Gold coated glass slide,
layer thickness 100 nm , gold, size 75 mm × 25 mm × 1 mm
Expand
View Pricing