Merck
  • Selective photo-reduction of p-nitrophenol to p-aminophenol by Au deposited CdS nanostructures of different shapes having large surface area.
Selective photo-reduction of p-nitrophenol to p-aminophenol by Au deposited CdS nanostructures of different shapes having large surface area.
Journal of nanoscience and nanotechnology (2013-08-02)
Nidhi Gupta, Bonamali Pal
ABSTRACT
The preparation and photocatalytic reductive ability of a new spongy flower or cotton bud like amorphous CdS microstructures (size = 200-300 nm) having intermediate crystal phase between cubic and hexagonal structure and possesses much larger surface area ca. 119 m2 g(-1) is demonstrated here. Structural analysis has been done by TEM, XRD, BET surface area, UV-Vis absorption, luminescence analysis, and photocatalytic study. The photoreactivity of as-prepared CdS for p-nitrophenol reduction to p-aminophenol conversion under visible light (150 W halogen lamp) irradiation has been found to be greatly enhanced from 62% to 94% (yield) and 81% to 100% (selectivity) after 1 wt% Au deposition. The obtained p-aminophenol yield is always better than 32% for CdS nanosphere (size 10-12 nm) and 40% for CdS nanorod (length/width in nm = 126/6). Surface passivation with a thin layer of SiO2 coating over this flower like spongy CdS appreciably increased the BET surface area to 158 m2 g(-1) and displayed stable photocatalytic activity for p-nitrophenol reduction up to 30 h of light exposure as compared to bare CdS of low activity because of its easy photodissolution. A good co-relation between the BET surface area and surface photoactivity of as-prepared CdS with the conventional cubic and hexagonal CdS nanoparticles of different dimensions is established here.
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), diam. × thickness 15 mm × 130-170 μm, layer thickness 100 Å
Sigma-Aldrich
Gold coated glass cover slip, L × W × thickness 22 mm × 22 mm × 130-170 μm, square, 99.999% (Au), layer thickness 100 Å
Sigma-Aldrich
Gold coated glass slide, layer thickness 1000 Å, 99.999% (Au)
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Cadmium selenide, -325 Mesh particle size, 99.99% trace metals basis, electronic grade
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold coated glass slide, 99.999% (Au), layer thickness 100 Å