Merck
  • Thickness dependence of the crystallization of Au/glass ultrathin films.
Thickness dependence of the crystallization of Au/glass ultrathin films.
Journal of nanoscience and nanotechnology (2013-07-19)
Tae-Sik Cho, Jin-Woo Kim
ABSTRACT
The crystallization of Au/glass ultrathin films for surface plasmon resonance (SPR) biosensor has been studied using synchrotron X-ray scattering and field emission scanning electron microscope. In films thinner than 30 nm, crystallized Au grains with [111] preferred orientation were formed in the as-deposited amorphous precursor. In film with 58-nm thickness, however, Au (200) powder grains existed on top of the Au (111)-oriented grains near the interface. At the annealing temperature of 400 degrees C, the Au (200) powder grains disappeared, while the Au (111)-oriented grains grew further. The behavior of the surface morphology of Au thin films post-annealed at 400 degrees C was consistent with the thickness dependence of the crystallization. In a 10-nm-thick film, the Au (111)-oriented grains fully crystallized, and then became separated with each other. By increasing to 30-nm film thickness, the Au (111)-oriented grains grew further, coalesced into large columnar-type grains, and showed smooth surface. We suggest that the appropriate thickness of Au/glass thin film for SPR biosensor need over 30-nm-thick, considering smooth surface.
Sigma-Aldrich
Gold, beads, 1-6 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.997% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.05 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, rod, diam. 3.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <45 μm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <10 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold coated glass slide, 99.999% (Au), layer thickness 1000 Å
Sigma-Aldrich
Gold coated glass slide, 99.999% (Au), layer thickness 100 Å
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), layer thickness 100 Å, diam. × thickness 15 mm × 130-170 μm
Sigma-Aldrich
Gold coated glass cover slip, 99.999% (Au), layer thickness 100 Å, L × W × thickness 22 mm × 22 mm × 130-170 μm, square
Sigma-Aldrich
Gold, nanopowder, <100 nm particle size, 99.9% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.1 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, evaporation slug, diam. × L 0.3 cm × 0.6 cm, 99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.025 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, powder, <850 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Gold, foil, thickness 0.127 mm, 99.99% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 0.5 mm, 99.999% trace metals basis
Sigma-Aldrich
Gold, wire, diam. 1.0 mm, 99.99% trace metals basis