Skip to Content
Merck
All Photos(2)

Documents

703192

Sigma-Aldrich

Indium tin oxide coated glass slide, square

surface resistivity 8-12 Ω/sq

Synonym(s):

ITO, ITO coated slide, square

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In2O3 · (SnO2)x
CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

description

slide

surface resistivity

8-12 Ω/sq

L × W × thickness

25 mm × 25 mm × 1.1 mm

transmittance

84% (nominal at 550nm)

refractive index

n20/D 1.517

SMILES string

O=[Sn]=O.O=[In]O[In]=O

InChI

1S/2In.5O.Sn

InChI key

LNNWKAUHKIHCKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium tin oxide (ITO) is widely used as transparent conducting oxides. Its major characteristics are electrical conductivity, optical transparency and the ease to be deposited as films.
substrate: unpolished float glass, SiO2 passivated (200-300 Å)

Application

ITO glass slides were used as a substrate for fabricating polymer solar cells based on blends of polymeric semiconductors and organic light emitting diodes (OLEDS). ITO substrates can also be used to design transparent heater arrays by etching the surface of the substrate by a printing tool. ITO cover slides may be used for closed digital microfluidic devices and as transparent electrodes in display and other optoelectronic devices.Some other probable uses that has been reported are:
  • Eumelanin samples prepared on ITO covered glass substrates.
  • Interface windows of the liquid cell for optical beam deflection (OBD) sensing contains a 20-nm-thick indium tin oxide film.
  • Mesoporous TiO2 films were formed on ITO conductive glasses.

Physical properties

Thickness of ITO coating is 1,200-1,600 Å

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keith A Brown et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(32), 12921-12924 (2013-07-19)
Scanning probe lithography (SPL) is a promising candidate approach for desktop nanofabrication, but trade-offs in throughput, cost, and resolution have limited its application. The recent development of cantilever-free scanning probe arrays has allowed researchers to define nanoscale patterns in a
Myungjae Yang et al.
Small (Weinheim an der Bergstrasse, Germany), 14(25), e1800885-e1800885 (2018-05-29)
A method is developed to directly map nanoscale "noise-source switching" phenomena during the optoelectronic switching of phase-separated polymer nanocomposites of tetrathiafulvalene (TTF) and phenyl-C61 -butyric acid methyl ester (PCBM) molecules dispersed in a polystyrene (PS) matrix. In the method, electrical
Online coupling of digital microfluidic devices with mass spectrometry detection using an eductor with electrospray ionization.
Baker CA and Roper MG
Analytical Chemistry, 2955-2960 (2012)
Preparation of non-annealed anatase TiO< sub> 2</sub> film on ITO substrate by anodizing in hot phosphate/glycerol electrolyte for dye-sensitized solar cells.
Tsuji E, et al.
Materials Letters, 91, 39-41 (2013)
Andreas Herrmann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(1), 117-124 (2008-12-17)
Application of an electric field to liquid crystalline film forming imines with negative dielectric anisotropy, such as N-(4-methoxybenzylidene)-4-butylaniline (MBBA, 1), results in the expulsion of compounds that do not participate in the formation of the liquid crystalline phase. Furthermore, amines

Articles

In this article, we demonstrate that bis-styrylbenzene derivatives show promising characteristics for very low lasing thresholds and discuss the design considerations for organic lasing molecules.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

Professor Shinar highlights low-cost, disposable sensor configurations in organic and hybrid electronics for healthcare applications.

See All

Protocols

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Selenium's roles range from essential dietary element to semiconductor in advanced applications like xerography.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service