Skip to Content
Merck
All Photos(1)

Key Documents

391956

Sigma-Aldrich

Dimethylamine solution

2.0 M in THF

Synonym(s):

N,N-Dimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH
CAS Number:
Molecular Weight:
45.08
Beilstein:
605257
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

liquid

concentration

1.90-2.20 M (by NaOH, titration)
2.0 M in THF

bp

~60 °C

density

0.85 g/mL at 25 °C

functional group

amine

SMILES string

CNC

InChI

1S/C2H7N/c1-3-2/h3H,1-2H3

InChI key

ROSDSFDQCJNGOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethylamine solution (2M in THF) has been used in the synthesis of 2-dimethylamino-1-(3,4-methylenedioxyphenyl)propan-1-one (bk-MDDMA) , a β-keto derivative of 3,4-methylenedioxyamphetamine (MDA) by reacting with 2-bromo-3′,4′-methylenedioxypropiophenone.[1] It may also be used to synthesize organic intermediates such as dimethyl-(4-nitro-benzyl)-amine, dimethyl-(4-methyl-benzyl)-amine and 4-dimethylamino-but-2-enoic acid [4-(3,4-dichloro-6-fluoro-phenylamino)-quinazolin-6-yl]-amide.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-32.8 °F - closed cup

Flash Point(C)

-36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[11C]-dimethylamine as a labeling agent for PET biomarkers.
Jacobson O & Mishani E.
Applied Radiation and Isotopes, 66(2), 188-193 (2008)
Discrimination and identification of regioisomeric ?-keto analogues of 3, 4-methylenedioxyamphetamines by gas chromatography-mass spectrometry.
Zaitsu K, et al.
Forensic Toxicology, 26(2), 45-51 (2008)
L Lee et al.
Cancer research, 41(10), 3992-3994 (1981-10-01)
Using a method for nitrosamine analysis that gives high recovery values and that is free from artifactual synthesis of nitrosamines, we have shown that human feces do not contain volatile nitrosamines (detection limit, 0.1 to 0.5 microgram/kg). We also showed
C Mulder et al.
Journal of neural transmission (Vienna, Austria : 1996), 109(9), 1203-1208 (2002-08-31)
Nitric oxide (NO) may play a role in the pathophysiology of Alzheimer's disease (AD). Asymmetric dimethylarginine (ADMA), an endogenous inhibitor of NO synthase, is involved in regulation of NO production. Recently it has been reported that dimethylarginine dimethylaminohydrolase, an enzyme
George Nicholson et al.
PLoS genetics, 7(9), e1002270-e1002270 (2011-09-21)
We have performed a metabolite quantitative trait locus (mQTL) study of the (1)H nuclear magnetic resonance spectroscopy ((1)H NMR) metabolome in humans, building on recent targeted knowledge of genetic drivers of metabolic regulation. Urine and plasma samples were collected from

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service