Skip to Content
Merck
All Photos(3)

Key Documents

230707

Sigma-Aldrich

n-Butyllithium solution

2.5 M in hexanes

Synonym(s):

n-BuLi, Butyl lithium, Butyllithium solution, Lithium-1-butanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Li
CAS Number:
Molecular Weight:
64.06
Beilstein:
1209227
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

2.5 M in hexanes

density

0.693 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li]CCCC

InChI

1S/C4H9.Li/c1-3-4-2;/h1,3-4H2,2H3;

InChI key

MZRVEZGGRBJDDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

n-Butyllithium (n-BuLi) is an organolithium reagent commonly used as a strong base in organic synthesis. It is also used as a lithium source for a wide range of lithium bases, such as lithium amides, acetylides, and alkoxides.

Application

n-BuLi can be used as a strong base to form corresponding lithium salts by the deprotonation of nitrogen, oxygen, phosphorus, and carbon acids. Heterocyclic compounds, such as furans, thiophenes, oxazoles, pyrroles, etc, can be lithiated α to the ring heteroatom using n-BuLi. These lithiated salts react in situ with alkyl halides to obtain useful organic compounds by the formation of the C-C bond. n-BuLi is also a useful reagent for lithium–halogen exchange reactions. Synthesis of various other useful reagents such as lithium diisopropylamide (LDA) and diphenylphosphine is done by in situ reaction with n-BuLi In the polymerization of dienes, n-BuLi is employed as an initiator.
The product is also used in the following reactions:      
  • Anionic rearrangement reactions      
  • Metal-halogen interchange and transmetalation reactions
  • Elimination reactions      
  • [1,2]- and [1,4]-Wittig rearrangement reaction      
  • Anionic homo-Fries rearrangement reaction      
  • Asymmetric carbolithiation

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

n-Butyllithium
Ovaska TV, et al.
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Microstructure-thermal property relationship of high trans-1, 4-poly (butadiene) produced by anionic polymerization of 1, 3-butadiene using an initiator composed of alkyl aluminum, n-butyl lithium, and barium alkoxide
Benvenuta-Tapia JJ, et al.
Polym. Eng. Sci., 49(1), 1-10 (2009)
Rhodium pincer complexes of 2, 2?-bis (diphenylphosphino) diphenylamine
Winter AM, et al.
Journal of Organometallic Chemistry, 682(1-2), 149-154 (2003)
Jennifer L Rutherford et al.
Journal of the American Chemical Society, 124(2), 264-271 (2002-01-10)
6Li and (13)C NMR spectroscopic studies were carried out on [(6)Li]n-BuLi and [(6)Li]PhLi (RLi) in toluene-d(8) containing the following diamines: N,N,N',N'-tetramethylethylenediamine (TMEDA), N,N,N',N'-tetraethylethylenediamine, 1,2-dipyrrolidinoethane, 1,2-dipiperidinoethane, N,N,N',N'-tetramethylpropanediamine, trans-(R,R)-N,N,N',N'-tetramethylcyclohexanediamine, and (-)-sparteine. Dimers of general structure (RLi)(2)S(2) (S = chelating diamine) are formed
Michael A Tarselli et al.
Organic letters, 11(20), 4596-4599 (2009-10-09)
A procedure for the coupling of aliphatic imines with allylic and allenic alkoxides is described. The success of these studies was enabled by a unique reactivity profile of Ti(IV) isopropoxide/n-BuLi compared to well-known Ti(IV) isopropoxide/RMgX systems.

Articles

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Transformative reagents enable selective conversions within molecules containing sensitive functionalities under mild reactions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service