Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Dimethylamine solution
Linear Formula:
(CH
3
)
2
NH
CAS Number:
124-40-3
Molecular Weight:
45.08
Products
Product Category
organic bases (4)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Quality Segment
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Purum (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
426458
Dimethylamine solution,
40 wt. % in H
2
O
Expand
View Pricing
391956
Dimethylamine solution,
2.0 M in THF
Expand
View Pricing
388246
Dimethylamine solution,
2.0 M in methanol
Expand
View Pricing
38950
Dimethylamine solution,
purum, 33% in absolute ethanol (~5.6 M)
Expand
View Pricing
1211200
Dimethylamine solution,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing