Skip to Content
Merck
All Photos(1)

Key Documents

295280

Sigma-Aldrich

Dimethylamine

anhydrous, ≥99%

Synonym(s):

N,N-Dimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NH
CAS Number:
Molecular Weight:
45.08
Beilstein:
605257
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22

grade

anhydrous

vapor density

1.55 (15 °C, vs air)

vapor pressure

1277 mmHg ( 20 °C)

Assay

≥99%

form

gas
liquid

autoignition temp.

753 °F

expl. lim.

14.4 %

bp

7 °C (lit.)

mp

−93 °C (lit.)

density

0.68 g/mL at 20 °C (lit.)

functional group

amine

SMILES string

CNC

InChI

1S/C2H7N/c1-3-2/h3H,1-2H3

InChI key

ROSDSFDQCJNGOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dimethylamine can be used:
  • In the preparation of 1-organyl-1-(trimethylsiloxy)-2-(dimethylamino)ethenes starting from α-bromo ketones.
  • In one of the key synthetic steps for the preparation of 1-methoxycarbonylpyrrolizin-3-one and its associated compounds.
  • Ni-catalyzed cross-electrophile coupling of Katritzky pyridinium salts with bromoalkanes through C-N bond activation.
  • To synthesize tertiary amides which on reaction with Petasis reagent followed by bromination forms bromomethyl ketones.

Packaging

Supplied in a Sure/Pac cylinder and has a 316 stainless steel needle valve with a male 1/4" NPTF outlet thread installed . Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

19.9 °F - closed cup

Flash Point(C)

-6.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bromination of Enamines from Tertiary Amides Using the Petasis Reagent: A Convenient One-Pot Regioselective Route to Bromomethyl Ketones
Kobeissi M, et al.
Synthetic Communications, 43(21), 2955-2965 (2013)
Ni-catalyzed deaminative cross-electrophile coupling of Katritzky salts with halides via C-N bond activation
Ni S, et al.
Science advances, 5(6), 9516-9516 (2019)
1-Methoxycarbonylpyrrolizin-3-one and related compounds
Despinoy XLM and McNab H
Organic & Biomolecular Chemistry, 7(10), 2187-2194 (2009)
Synthesis of 1-organyl-1-(trimethylsiloxy)-2-(dimethylamino) ethenes and new hypervalent silicon compounds based thereon
Gostevskii BA, et al.
Russ. J. Org. Chem., 52(10), 1385-1389 (2016)
L Lee et al.
Cancer research, 41(10), 3992-3994 (1981-10-01)
Using a method for nitrosamine analysis that gives high recovery values and that is free from artifactual synthesis of nitrosamines, we have shown that human feces do not contain volatile nitrosamines (detection limit, 0.1 to 0.5 microgram/kg). We also showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service