Skip to Content
Merck
All Photos(1)

Documents

251682

Sigma-Aldrich

Sodium 3-mercapto-1-propanesulfonate

technical grade, 90%

Synonym(s):

3-Mercapto-1-propanesulfonic acid sodium salt, MPS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2CH2SO3Na
CAS Number:
Molecular Weight:
178.21
Beilstein:
5362242
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

90%

form

powder

mp

~220 °C (dec.) (lit.)

solubility

water: soluble 50 mg/mL, colorless

SMILES string

[Na+].[O-]S(=O)(=O)CCCS

InChI

1S/C3H8O3S2.Na/c4-8(5,6)3-1-2-7;/h7H,1-3H2,(H,4,5,6);/q;+1/p-1

InChI key

FRTIVUOKBXDGPD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 3-mercapto-1-propanesulfonate forms self assembled monolayer via chemisorption on a gold-coated substrate.

Application

Sodium 3-mercapto-1-propanesulfonate (MPS) can be used to prepare tetrasubstituted organosilicon thioether by photochemical reaction with tetravinylsilane via thiol-ene reaction. It can also be used as a capping agent to functionalize gold nanoparticles for potential applications in the field of radiotherapy and drug delivery.
MPS can be also used in the preparation of Rh nanoparticles by liquid-phase reduction of rhodium (III) chloride.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thiol- Ene Reaction for the Synthesis of Multifunctional Branched Organosilanes
Rissing C and Son DY
Organometallics, 27(20), 5394-5397 (2008)
Geoffrey J Ashwell et al.
Faraday discussions, 131, 23-31 (2006-03-04)
The chevron-shaped dye, N-butyl-2,6-bis-[2-{4-(2-(4-dibutylaminophenyl)-vinyl)-phenyl}-vinyl]-pyridinium iodide, has a hydrophilic apex that is electron-accepting and hydrophobic it-bridged limbs that are electron-donating. It forms non-centrosymmetric monolayers at the air-water interface and when deposited, its LB films exhibit second-harmonic generation and asymmetric current-voltage (I-V)
J Tie et al.
Annals of oncology : official journal of the European Society for Medical Oncology, 26(8), 1715-1722 (2015-04-09)
Early indicators of treatment response in metastatic colorectal cancer (mCRC) could conceivably be used to optimize treatment. We explored early changes in circulating tumor DNA (ctDNA) levels as a marker of therapeutic efficacy. This prospective study involved 53 mCRC patients
Highly dispersed noble-metal/chromia (core/shell) nanoparticles as efficient hydrogen evolution promoters for photocatalytic overall water splitting under visible light
Sakamoto N, et al.
Nanoscale, 1(1), 106-109 (2009)
Nucleobases functionalized quantum dots and gold nanoparticles bioconjugates as a fluorescence resonance energy transfer (FRET) system-Synthesis, characterization and potential applications
Rodzik-Czalka L, et al.
Journal of Colloid and Interface Science, 514(20), 479-490 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service