Skip to Content
Merck
All Photos(1)

Key Documents

M1511

Supelco

Sodium 2-mercaptoethanesulfonate

analytical standard, ≥98.0% (titration)

Synonym(s):

2-Mercaptoethanesulfonic acid sodium salt, Coenzyme M sodium salt, HS-CoM Na, MESNA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2SO3Na
CAS Number:
Molecular Weight:
164.18
Beilstein:
3657828
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (titration)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

[Na+].[O-]S(=O)(=O)CCS

InChI

1S/C2H6O3S2.Na/c3-7(4,5)2-1-6;/h6H,1-2H2,(H,3,4,5);/q;+1/p-1

InChI key

XOGTZOOQQBDUSI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 2-mercaptoethanesulfonate is a thiol compound commonly used as an auxiliary drug, to reduce the side effects of anti-cancer drugs, such as oxazaphosphorine, cyclophosphamide, and ifosfamide. It acts by forming a non-toxic compound upon reacting with the toxic metabolites of the drugs like acrolein.

Application

The analytical standard can also be used as follows:
  • Quantification of sodium 2-mercaptoethanesulfonate (MESNA) from human samples by micellar electrokinetic chromatography (MEKC) coupled with UV detection and in-capillary derivatization of MESNA
  • Development of a fluorescence-enhanced histidine stabilized gold nanoclusters (His-Au NCs) based probe for the detection of sodium 2-mercaptoethanesulfonate in uromitexan samples
  • High-performance liquid chromatography-tandem mass spectrometric based bioanalysis of sodium 2-mercaptoethanesulfonate in rat plasma samples
  • To quantify sodium 2-mercaptoethanesulfonate in two of its pharmaceutical preparations (injections) by surface-enhanced Raman scattering (SERS) using gold colloid as a probe

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John H Lewis et al.
Nature structural & molecular biology, 26(9), 802-807 (2019-09-07)
Conformational changes within typical protein molecules are rapid and small, making their quantitative resolution challenging. These changes generally involve rotational motions and may thus be resolved by determining changes in the orientation of a fluorescent label that assumes a unique
Muhammad Faizan Nazar et al.
Journal of colloid and interface science, 363(2), 490-496 (2011-08-27)
A viable cost-effective approach employing mixtures of non-ionic surfactants Triton X-114/Triton X-100 (TX-114/TX-100), and subsequent cloud point extraction (CPE), has been utilized to concentrate and recycle inorganic nanoparticles (NPs) in aqueous media. Gold Au- and palladium Pd-NPs have been pre-synthesized
Twalib Ngoma et al.
British journal of haematology, 158(6), 749-762 (2012-08-01)
Prior to the introduction of the International Network for Cancer Treatment and Research (INCTR) protocol INCTR 03-06, survival of patients with Burkitt lymphoma at four tertiary care centres in equatorial Africa was probably no more than 10-20%. The results reported
Jun-ichi Nishigaki et al.
Inorganic chemistry, 51(6), 3690-3697 (2012-03-10)
Divalent and trivalent nickel complexes of 1,4,8,11-tetraazacyclotetradecane, denoted as cyclam hereafter, coordinated by methyl coenzyme M (MeSCoM(-)) and coenzyme M (HSCoM(-)) have been synthesized in the course our model studies of methyl coenzyme M reductase (MCR). The divalent nickel complexes
Astrid Hoeppner et al.
Acta crystallographica. Section D, Biological crystallography, 68(Pt 11), 1549-1557 (2012-10-24)
The zinc-containing corrinoid:coenzyme M methyltransferase MtaA is part of the methanol-coenzyme M-methyltransferase complex of Methanosarcina mazei. The whole complex consists of three subunits: MtaA, MtaB and MtaC. The MtaB-MtaC complex catalyses the cleavage of methanol (bound to MtaB) and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service