63707
2-Mercaptoethanesulfonic acid solution
for protein sequence analysis, ampule, 3.0 M±0.1 M in H2O (T)
Synonym(s):
β-Mercaptoethanesulfonic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
for protein sequence analysis
quality
ampule
concentration
3.0 M±0.1 M in H2O (T)
density
1.25 g/mL at 20 °C
SMILES string
OS(=O)(=O)CCS
InChI
1S/C2H6O3S2/c3-7(4,5)2-1-6/h6H,1-2H2,(H,3,4,5)
InChI key
ZNEWHQLOPFWXOF-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Environmental pollution (Barking, Essex : 1987), 250, 233-240 (2019-04-19)
Microbes in sediments contribute to nutrient release and play an important role in lake eutrophication. However, information about the profiles of functional genes and bacterial communities and the most important environmental factor affecting them in the sediments of eutrophic lake
Microbial cell factories, 14, 33-33 (2015-04-17)
Anaerobic digestion has been widely applied to treat the waste activated sludge from biological wastewater treatment and produce methane for biofuel, which has been one of the most efficient solutions to both energy crisis and environmental pollution challenges. Anaerobic digestion
Scientific reports, 9(1), 13847-13847 (2019-09-27)
The Guaymas Basin spreading center, at 2000 m depth in the Gulf of California, is overlain by a thick sedimentary cover. Across the basin, localized temperature anomalies, with active methane venting and seep fauna exist in response to magma emplacement
Nature communications, 11(1), 6381-6381 (2020-12-16)
A key feature that differentiates prokaryotic cells from eukaryotes is the absence of an intracellular membrane surrounding the chromosomal DNA. Here, we isolate a member of the ubiquitous, yet-to-be-cultivated phylum 'Candidatus Atribacteria' (also known as OP9) that has an intracytoplasmic
International journal of clinical oncology, 18(1), 170-176 (2011-12-20)
Good local control of high-grade non-small round cell soft tissue sarcomas (NSRCSTSs) has been achieved with significant advances in surgical techniques and radiotherapy. However, the role of chemotherapy remains controversial. Our aim was to investigate the efficacy, feasibility and adverse
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service