Skip to Content
Merck
All Photos(2)

Documents

375411

Sigma-Aldrich

Sodium 3-sulfobenzoate

97%

Synonym(s):

3-Carboxybenzenesulfonic acid sodium salt, 3-Sulfobenzoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO3SC6H4CO2H
CAS Number:
Molecular Weight:
224.17
Beilstein:
5675222
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

mp

≥300 °C (lit.)

solubility

water: soluble 25 mg/mL, clear, colorless

SMILES string

[Na+].OC(=O)c1cccc(c1)S([O-])(=O)=O

InChI

1S/C7H6O5S.Na/c8-7(9)5-2-1-3-6(4-5)13(10,11)12;/h1-4H,(H,8,9)(H,10,11,12);/q;+1/p-1

InChI key

KQHKITXZJDOIOD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium 3-sulfobenzoate is the sodium salt of 3-sulfobenzoic acid. Steady state method has been proposed for the evaluation of solubility of its binary solvent mixtures at high temperatures.

Application

Sodium 3-sulfobenzoate has been used in the synthesis of ionic liquid, 1-hexyl-3-propanenitrileimidazolium sulfobenzoate (SBA).
Sodium 3-sulfobenzoate is suitable for use in the preparation of {[Ce(3-SBH)(H2O)4]2CB6}·4NO3 and {[Nd(3-SBH)(H2O)4]2CB6}·4NO3 [SBH=sulfobenzoic acid, CB6=cucurbit[6]uril] complexes. It may be used in the preparation of 4,4′-(ethene-1,2-diyl)dipyridinium bis(3-carboxybenzenesulfonate).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Wu et al.
Acta crystallographica. Section E, Structure reports online, 70(Pt 11), o1170-o1170 (2014-12-09)
In the title mol-ecular salt, C12H12N2 (2+)·2C7H5O5S(-), the complete dication is generated by crystallographic inversion symmetry. In the anion, the sulfonic acid group is deprotonated and the dihedral angle between the planes of the carb-oxy-lic acid group and the benzene
Densities, viscosities, and refractive indices of 1-hexyl-3-propanenitrile imidazolium ionic liquids incorporated with sulfonate-based anions.
Ziyada AK, et al.
Journal of Chemical and Engineering Data, 56(5), 2343-2348 (2011)
Equilibrium solubility of sodium 3-sulfobenzoate in binary (sodium chloride+ water),(sodium sulfate+ water), and (ethanol+ water) solvent mixtures at temperatures from (278.15 to 323.15) K.
Li R, et al.
The Journal of Chemical Thermodynamics, 79, 8-11 (2014)
Lanthanide Ion Complexes with 2-, 3-, or 4-Sulfobenzoate and Cucurbit [6] uril.
Thue'ry P.
Crystal Growth & Design, 12(3), 1632-1640 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service