Skip to Content
Merck
All Photos(1)

Documents

108995

Sigma-Aldrich

Methyl thioglycolate

95%

Synonym(s):

Methyl mercaptoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CO2CH3
CAS Number:
Molecular Weight:
106.14
Beilstein:
506259
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

reaction suitability

reagent type: ligand
reaction type: [1,2]-Wittig Rearrangement

refractive index

n20/D 1.466 (lit.)

bp

42-43 °C/10 mmHg (lit.)

density

1.187 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CS

InChI

1S/C3H6O2S/c1-5-3(4)2-6/h6H,2H2,1H3

InChI key

MKIJJIMOAABWGF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl thioglycolate reacts with nonprotein component of the antitumor antibiotic neocarzinostatin to form 1:1 adduct. It reacts with isothiocyanate to form Rhodanine.

Application

Methyl thioglycolate was used in the preparation of:
  • 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene
  • methyl thioglycolate and aminoethanethiol conjugated gold nanorods.
Thiol Organocatalyst used in conjunction with visible light photocatalysis to affect the direct arylation of benzylic ether C-H bonds

A General Strategy for Organocatalytic Activation of C–H Bonds via Photoredox Catalysis: Direct Arylation of Benzylic Ethers

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

132.8 °F - closed cup

Flash Point(C)

56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Y Sugiura et al.
Biochemistry, 30(12), 2989-2992 (1991-03-26)
The reaction products of methyl thioglycolate with dynemicin A, dynemicin H and dynemicin S, were isolated by HPLC purification and identified spectroscopically. The major product, dynemicin H (C30H23NO9), was determined to be a C-8 hydrogen analogue of dynemicins L and
W A LaMarr et al.
Proceedings of the National Academy of Sciences of the United States of America, 95(1), 102-107 (1998-02-21)
DNA superhelical tension, an important feature of genomic organization, is known to affect the interactions of intercalating molecules with DNA. However, the effect of torsional tension on nonintercalative DNA-binding chemicals has received less attention. We demonstrate here that the enediyne
M K Vadnere et al.
Journal of medicinal chemistry, 29(9), 1714-1720 (1986-09-01)
The reaction of methyl mercaptoacetate (5) with phenyl-p-benzoquinone (6) or 5-p-benzoquinonyl-3',5'-di-O-acetyl-2'-deoxyuridine (10) resulted in the formation of the three possible adducts to the quinone rings of 6 and 10; an additional product in the reaction with 10 was the unsubstituted
D J Creighton et al.
Biochemistry, 27(19), 7376-7384 (1988-09-20)
A quantitative kinetic model for the glutathione-dependent conversion of methylglyoxal to D-lactate in mammalian erythrocytes has been formulated, on the basis of the measured or calculated rate and equilibrium constants associated with (a) the hydration of methylglyoxal, (b) the specific
Stereochemical assignment of neocarzinostatin chromophore. Structures of neocarzinostatin chromophore-methyl thioglycolate adducts.
Myers AG, et al.
Journal of the American Chemical Society, 110(21), 7212-7214 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service