Skip to Content
Merck
All Photos(2)

Key Documents

171476

Sigma-Aldrich

Undecanoic acid

98%

Synonym(s):

Hendecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9COOH
CAS Number:
Molecular Weight:
186.29
Beilstein:
1759287
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

bp

228 °C/160 mmHg (lit.)
248-250 °C (lit.)

mp

28-31 °C (lit.)

functional group

carboxylic acid

SMILES string

CCCCCCCCCCC(O)=O

InChI

1S/C11H22O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3,(H,12,13)

InChI key

ZDPHROOEEOARMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Undecanoic acid was used in the formation of cycloamylose crystals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>233.6 °F

Flash Point(C)

> 112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rashanique D Quarels et al.
Beilstein journal of nanotechnology, 8, 1863-1877 (2017-10-20)
Visible-light irradiation of phthalimide esters in the presence of the photosensitizer [Ru(bpy)
Nicole Lindenkamp et al.
Applied and environmental microbiology, 78(15), 5375-5383 (2012-05-29)
β-Ketothiolases catalyze the first step of poly(3-hydroxybutyrate) [poly(3HB)] biosynthesis in bacteria by condensation of two acetyl coenzyme A (acetyl-CoA) molecules to acetoacetyl-CoA and also take part in the degradation of fatty acids. During growth on propionate or valerate, Ralstonia eutropha
Margarida M L M Vareiro et al.
Analytical biochemistry, 377(2), 243-250 (2008-04-03)
The development of a single-step, separation-free method for measurement of low concentrations of fatty acid using a surface plasmon resonance-enhanced fluorescence competition assay with a surface-bound antibody is described. The assay behavior was unexpectedly complex. A nonlinear coverage-dependent self-quenching of
Fernanda G Paião et al.
FEMS microbiology letters, 271(2), 180-186 (2007-04-12)
Suppressive subtractive hybridization was used to isolate transcripts specifically upregulated during Trichophyton rubrum exposure to acriflavin, fluconazole, griseofulvin, terbinafine or undecanoic acid. Macro-array dot-blot and sequencing of 132 clones, which correspond to genes differentially expressed after exposition of T. rubrum
Dorit Avrahami et al.
Biochemistry, 42(50), 14946-14956 (2003-12-17)
The dramatically increased frequency of opportunistic fungal infections has prompted research to diversify the arsenal of antifungal agents. Antimicrobial peptides constitute a promising family for future antibiotics with a new mode of action. However, only a few are effective against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service