Skip to Content
Merck
All Photos(1)

Key Documents

T0502

Sigma-Aldrich

Tridecanoic acid

≥98%

Synonym(s):

Tridecylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11CO2H
CAS Number:
Molecular Weight:
214.34
Beilstein:
508317
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥98%

form

(powder, crystals or flakes)

bp

236 °C/100 mmHg (lit.)

mp

41-42 °C (lit.)

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC(O)=O

InChI

1S/C13H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13(14)15/h2-12H2,1H3,(H,14,15)

InChI key

SZHOJFHSIKHZHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Tridecanoic acid is an aliphatic monoacid. It is complexed to β-cyclodextrin (β-CD) for structural studies. It can exist in three polymeric forms(A′, B′ and C′) when crystallized.

Application

Tridecanoic acid has been used as an internal standard for the lipid analysis of total intramuscular fat from broilers and fatty acid analysis in chinook salmon roe lipids. It has also been used as a reference standard in gas chromatography for quantifying fatty acids in meat samples.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Echium oil is better than rapeseed oil in enriching poultry meat with n-3 polyunsaturated fatty acids, including eicosapentaenoic acid and docosapentaenoic acid
Kitessa SM and Young P
The British Journal of Nutrition, 101(5), 709-715 (2008)
The Crystal Structure of the A?-form of Tridecanoic Acid
Goto M and Asada E
Bulletin of the Chemical Society of Japan, 53(8), 2111-2113 (1980)
Effects of dietary oregano essential oil, laurel essential oil and attapulgite on chemical composition, oxidative stability, fatty acid profile and mineral content of chicken breast and thigh meat
Giannenas I, et al.
European Poultry Science, 80, 318-325 (2016)
Gabriela Christina Kuhl et al.
Microbiology spectrum, 9(2), e0117921-e0117921 (2021-10-14)
Conjugated linoleic acid (CLA) has been the subject of numerous studies in recent decades because of its associated health benefits. CLA is an intermediate product of the biohydrogenation pathway of linoleic acid (LA) in bacteria. Several bacterial species capable of
Impact of maturity on the physicochemical and biochemical properties of chinook salmon roe
Bekhit AEDA, et al.
Food Chemistry, 117(2), 318-325 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service