Skip to Content
Merck
All Photos(1)

Key Documents

89764

Supelco

Undecanoic acid

analytical standard

Synonym(s):

Hendecanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9COOH
CAS Number:
Molecular Weight:
186.29
Beilstein:
1759287
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

228 °C/160 mmHg (lit.)
248-250 °C (lit.)

mp

28-31 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCC(O)=O

InChI

1S/C11H22O2/c1-2-3-4-5-6-7-8-9-10-11(12)13/h2-10H2,1H3,(H,12,13)

InChI key

ZDPHROOEEOARMN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Undecanoic acid is a medium chain fatty acid and the most fungitoxic compound commonly used in the treatment of superficial mycoses in humans.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Undecanoic acid may be used as an internal standard for the determination of medium-chain fatty acids in human fecal water, fecal fermentation supernatants, and human plasma by gas chromatography-mass spectrometry method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>233.6 °F

Flash Point(C)

> 112 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transcriptome-wide survey of gene expression changes and alternative splicing in Trichophyton rubrum in response to undecanoic acid.
Mendes NS, et al.
Scientific Reports, 8(1), 2520-2520 (2018)
Development of a fast and cost-effective gas chromatography- mass spectrometry method for the quantification of short-chain and medium-chain fatty acids in human biofluids.
Lotti C, et al.
Analytical and Bioanalytical Chemistry, 409(23), 5555-5567 (2017)
Joseph J Pesek et al.
Journal of chromatography. A, 1216(7), 1140-1146 (2009-01-13)
The use of silica hydride-based stationary phases for the retention and analysis of nucleotides has been investigated. Both reversed-phase columns with a hydride surface underneath as well as those with an unmodified or a minimally modified hydride material were tested.
Jorge Almodóvar et al.
Biotechnology and bioengineering, 110(2), 609-618 (2012-08-21)
Cortical bone allografts suffer from high rates of failure due to poor integration with host tissue, leading to non-union, fracture, and infection following secondary procedures. Here, we report a method for modifying the surfaces of cortical bone with coatings that
Hiromi Kitano et al.
Journal of colloid and interface science, 250(1), 134-141 (2005-11-18)
A disulfide, which carried two L-phenylalanyl p-nitroanilide (Phe-pNA) moieties at both ends, was prepared by the coupling of 11,11'-dithiodiundecanoic acid (DTUA) with Phe-pNA. The compound obtained (DTUA-Phe-pNA) formed a self-assembled monolayer (SAM) on a gold electrode and vacuum-evaporated gold thin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service