Skip to Content
Merck
All Photos(1)

Key Documents

N5502

Sigma-Aldrich

Nonanoic acid

≥97%

Synonym(s):

Acid C9, Pelargonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7COOH
CAS Number:
Molecular Weight:
158.24
Beilstein:
1752351
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

vapor density

5.5 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥97%

form

liquid

expl. lim.

9 %

refractive index

n20/D 1.432 (lit.)

bp

268-269 °C (lit.)

mp

9 °C (lit.)

density

0.906 g/mL at 25 °C (lit.)

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CCCCCCCCC(O)=O

InChI

1S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)

InChI key

FBUKVWPVBMHYJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Qualitative and Quantitative Metabolite Comparison of Korean Traditional Alcoholic Beverages: Takju, Yakju, and Traditional-Soju.: Uses nonanoic acid to analyze and compare the metabolic profiles of traditional Korean alcoholic beverages, aiding in the preservation of cultural heritage and improving product quality (Tak et al., 2024).

  • In Vitro Evaluation of Essential Oils and Saturated Fatty Acids for Repellency against the Old-World Sand Fly, Phlebotomus papatasi (Scopoli) (Diptera: Psychodidae).: This research explores nonanoic acid′s potential as a natural repellent, contributing to the development of safer and more effective control measures against vector-borne diseases (Temeyer et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

278.6 °F - Pensky-Martens closed cup

Flash Point(C)

137 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Hoschek et al.
Biotechnology and bioengineering, 116(8), 1887-1900 (2019-05-01)
Photoautotrophic organisms are promising hosts for biocatalytic oxyfunctionalizations because they supply reduction equivalents as well as O2 via photosynthetic water oxidation. Thus far, research on photosynthesis-driven bioprocesses mainly focuses on strain development and the proof of principle in small-scale biocatalytic
Anders Clemmensen et al.
The Journal of investigative dermatology, 130(9), 2201-2210 (2010-04-30)
The pathogenesis of irritant contact dermatitis (ICD) is poorly understood, and genes participating in the epidermal response to chemical irritants are only partly known. It is commonly accepted that different irritants have different mechanisms of action in the development of
Zhiyong Sun et al.
Applied microbiology and biotechnology, 74(1), 69-77 (2006-10-26)
Pseudomonas putida KT2440 grew on glucose at a specific rate of 0.48 h(-1) but accumulated almost no poly-3-hydroxyalkanoates (PHA). Subsequent nitrogen limitation on nonanoic acid resulted in the accumulation of only 27% medium-chain-length PHA (MCL-PHA). In contrast, exponential nonanoic acid-limited
Zhiyong Sun et al.
Journal of biotechnology, 143(4), 262-267 (2009-07-28)
Medium-chain-length polyhydroxyalkanoates (MCL-PHAs) were produced in carbon-limited, single-stage, fed-batch fermentations of Pseudomonas putida KT2440 by co-feeding nonanoic acid (NA) and glucose (G) to enhance the yield of PHA from NA. An exponential (mu=0.25 h(-1)) followed by a linear feeding strategy
Flemming Andersen et al.
Contact dermatitis, 55(3), 155-159 (2006-08-22)
So-called anti-irritants (AI) are widely used in cosmetic formulations, with the aim of reducing irritation from substances in the formulation. It may also be claimed that they are 'soothing' and 'healing' ingredients. However, the proof for these claims is circumstantial.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service