Skip to Content
Merck
All Photos(3)

Documents

142794

Sigma-Aldrich

5-Sulfoisophthalic acid sodium salt

95%

Synonym(s):

5-Sodiosulfoisophthalic acid, 5-Sulfoisophthalic acid monosodium salt, Monosodium 5-sulfoisophthalate, Sodium 3,5-dicarboxybenzene-1-sulfonate, Sodium 5-sulfoisophthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaO3SC6H3-1,3-(CO2H)2
CAS Number:
Molecular Weight:
268.18
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

powder

SMILES string

[Na+].OC(=O)c1cc(cc(c1)S([O-])(=O)=O)C(O)=O

InChI

1S/C8H6O7S.Na/c9-7(10)4-1-5(8(11)12)3-6(2-4)16(13,14)15;/h1-3H,(H,9,10)(H,11,12)(H,13,14,15);/q;+1/p-1

InChI key

YXTFRJVQOWZDPP-UHFFFAOYSA-M

Application

5-Sulfoisophthalic acid sodium salt is a sulfonate carboxylate ligand which can be used in a variety of applications such as catalysis, gas storage, magnetic materials, and luminescence.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, Structure, Luminescent and Thermal Properties of Ytterbium (III) and Dysprosium (III) Complexes with 5-Sulfoisophthalic Acid Sodium Salt
Wu G, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 637(5), 602-607 (2011)
Cressa Ria P Fulong et al.
Dalton transactions (Cambridge, England : 2003), 47(24), 7905-7915 (2018-02-16)
Discrete metal-organic polyhedra (MOPs) containing copper(ii), palladium(ii), and iron(ii) nodes were synthesized as fillers for mixed-matrix materials (MMMs) with a polyvinylidine fluoride (PVDF) polymer phase and contrasted against an MMM containing a metal-organic framework, MOF-5. When a given MOP was
Synthesis, Structure, and Physical Properties of a Barium Complex with 5-Sulfoisophthalic Acid Sodium Salt
Wu G, et al.
Zeitschrift fur Anorganische und Allgemeine Chemie, 637(5), 596-601 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service